missing translation for 'onlineSavingsMsg'
Learn More

Iodine monobromide, 98%

CAS: 7789-33-5 | BrI | 206.81 g/mol

Supplier:  Thermo Scientific Chemicals A1615822

 View more versions of this product

Catalog No. AAA1615822


Only null left
Explore available promotions

Description

Description

Iodine monobromide is used as an electrophile employed in a new synthetic approach to polyketide structural units. It is also used in iodometry and serves as a source of I+. It is a powerful iodinating agent used in organic synthesis as well as involved in the synthesis of radioiodinated fatty acids for heart imaging.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iodine monobromide is used as an electrophile employed in a new synthetic approach to polyketide structural units. It is also used in iodometry and serves as a source of I+. It is a powerful iodinating agent used in organic synthesis as well as involved in the synthesis of radioiodinated fatty acids for heart imaging.

Solubility
Soluble in water, alcohol, ether, carbon disulfide and glacial acetic acid.

Notes
Air, light and moisture sensitive. Store in cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with alcohols, phosphorus, sodium and sodiumoxides and potassium.
Specifications

Specifications

116°C (decomposition)
50°C (sublimation)
100 g
MFCD00011353
3902973
14,5016
Soluble in water,alcohol,ether,carbon disulfide and glacial acetic acid.
BrI
206.81
206.81
Pungent
4.416
Iodine monobromide
7789-33-5
BrI
UN3260
None
iodine monobromide, iodine bromide, iodobromine, iodine bromide ibr, unii-g0k622rd9n, iodinemonobromide, iodobromane, bromoiodine, jodmonobromid, hanus solution
CBEQRNSPHCCXSH-UHFFFAOYSA-N
iodobromane
82238
98%
Light and moisture sensitive

Chemical Identifiers

7789-33-5
206.81
CBEQRNSPHCCXSH-UHFFFAOYSA-N
82238
BrI
BrI
MFCD00011353
iodine monobromide, iodine bromide, iodobromine, iodine bromide ibr, unii-g0k622rd9n, iodinemonobromide, iodobromane, bromoiodine, jodmonobromid, hanus solution
iodobromane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.

EINECSNumber : 232-159-9

TSCA : Yes

Recommended Storage : Keep cold; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only