missing translation for 'onlineSavingsMsg'
Learn More

Iodine (Iodine-Iodide), 0.0473 N, for Iodine Adsorption Number of Carbon Black, Ricca Chemical

Catalog No. 39901
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39901 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 39901 Supplier Ricca Chemical Company Supplier No. 39901

For Analysis of Iodine Adsorption Number of Carbon Black

Chemical Identifiers

CAS 7553-56-2
Molecular Formula I2
Molecular Weight (g/mol) 253.81
MDL Number MFCD00011355 MFCD00164163
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
Synonym iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
PubChem CID 807
ChEBI CHEBI:17606
IUPAC Name diiodine
SMILES II

Specifications

Grade Laboratory
Concentration or Composition (by Analyte or Components) 0.0473N
Packaging Amber Glass Bottle
Quantity 4 L
Chemical Name or Material Iodine (Iodine-Iodide) Solution