missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodine (Iodine-Iodide), 0.00500 N (N/200), Ricca Chemical
Supplier: Ricca Chemical Company R39705004C
Specifications
| Iodine | |
| Liquid | |
| I2 | |
| iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode | |
| PNDPGZBMCMUPRI-UHFFFAOYSA-N | |
| diiodine | |
| 807 | |
| Laboratory | |
| Amber Glass Bottle |
| 7732-18-5,7681-11-0,7553-56-2 | |
| 4 L | |
| MFCD00011355 MFCD00164163 | |
| Solubility in Water: Infinite | |
| II | |
| 253.81 | |
| CHEBI:17606 | |
| Characteristic weak Iodine odor | |
| Dark brown liquid |
Chemical Identifiers
| 7732-18-5 | |
| 253.81 | |
| PNDPGZBMCMUPRI-UHFFFAOYSA-N | |
| 807 | |
| diiodine |
| I2 | |
| MFCD00011355 MFCD00164163 | |
| iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode | |
| CHEBI:17606 | |
| II |
Safety and Handling
Recommended Storage : General