missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodine, Certified, 0.0200N ±0.0002N (0.01M), LabChem™
$78.47 - $253.67
Chemical Identifiers
| CAS | 7553-56-2 |
|---|---|
| Molecular Formula | I2 |
| Molecular Weight (g/mol) | 253.81 |
| MDL Number | MFCD00011355 MFCD00164163 |
| InChI Key | PNDPGZBMCMUPRI-UHFFFAOYSA-N |
| Synonym | iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode |
| PubChem CID | 807 |
| ChEBI | CHEBI:17606 |
| IUPAC Name | diiodine |
| SMILES | II |
Chemical Identifiers
| 7553-56-2 | |
| 253.81 | |
| PNDPGZBMCMUPRI-UHFFFAOYSA-N | |
| 807 | |
| diiodine |
| I2 | |
| MFCD00011355 MFCD00164163 | |
| iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode | |
| CHEBI:17606 | |
| II |
Specifications
| 7553-56-2,7732-18-5,7681-11-0,7647-01-0 | |
| 0.26,99.22,0.5,0.02 | |
| 1 L | |
| I2 | |
| iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode | |
| PNDPGZBMCMUPRI-UHFFFAOYSA-N | |
| diiodine | |
| 807 | |
| 253.809 | |
| 0.0200N ±0.0002N (0.01M) | |
| Amber Glass |
| Red through Brown | |
| Liquid | |
| I2 | |
| MFCD00011355 MFCD00164163 | |
| Soluble in water | |
| II | |
| 253.81 | |
| CHEBI:17606 | |
| Certified | |
| Passes Test | |
| Iodine |
Safety and Handling
GHS H Statement
Harmful to aquatic life.
GHS P Statement
Avoid release to the environment.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage : Room Temperature