missing translation for 'onlineSavingsMsg'
Learn More

Iodine, Certified, 0.0200N ±0.0002N (0.01M), LabChem™

Supplier:  LabChem LC156104

 View more versions of this product

Catalog No. LC156104


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Iodine
Red through Brown
Liquid
I2
MFCD00011355 MFCD00164163
Soluble in water
II
253.81
CHEBI:17606
Certified
Passes Test
7553-56-2,7732-18-5,7681-11-0,7647-01-0
0.26,99.22,0.5,0.02
4 L
I2
iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
PNDPGZBMCMUPRI-UHFFFAOYSA-N
diiodine
807
253.809
0.0200N ±0.0002N (0.01M)
Amber Glass

Chemical Identifiers

7553-56-2
253.81
PNDPGZBMCMUPRI-UHFFFAOYSA-N
807
diiodine
I2
MFCD00011355 MFCD00164163
iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
CHEBI:17606
II
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful to aquatic life.

GHS P Statement
Avoid release to the environment.
Dispose of contents/container to comply with local, state and federal regulations.

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions