missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodine, Certified, 0.0100N ±0.0001N (0.005M), LabChem™
$62.77 - $181.77
Chemical Identifiers
| CAS | 7553-56-2 |
|---|---|
| Molecular Formula | I2 |
| Molecular Weight (g/mol) | 253.81 |
| MDL Number | MFCD00011355 MFCD00164163 |
| InChI Key | PNDPGZBMCMUPRI-UHFFFAOYSA-N |
| PubChem CID | 807 |
| ChEBI | CHEBI:17606 |
| IUPAC Name | diiodine |
| SMILES | II |
Chemical Identifiers
| 7553-56-2 | |
| 253.81 | |
| PNDPGZBMCMUPRI-UHFFFAOYSA-N | |
| CHEBI:17606 | |
| II |
| I2 | |
| MFCD00011355 MFCD00164163 | |
| 807 | |
| diiodine |
Specifications
| 7553-56-2,7732-18-5,7681-11-0,7647-01-0 | |
| 0.13,99.6,0.25,0.02 | |
| 500 mL | |
| I2 | |
| iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode | |
| PNDPGZBMCMUPRI-UHFFFAOYSA-N | |
| diiodine | |
| 807 | |
| 253.809 | |
| 0.0100N ±0.0001N (0.005M) | |
| Amber Glass |
| Red through Brown | |
| Liquid | |
| I2 | |
| MFCD00011355 MFCD00164163 | |
| Soluble in water | |
| II | |
| 253.81 | |
| CHEBI:17606 | |
| Certified | |
| Passes Test | |
| Iodine |
Safety and Handling
GHS H Statement
Solution is not hazardous.
GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage : Room Temperature