missing translation for 'onlineSavingsMsg'
Learn More

Iodine, Certified, 0.0100N ±0.0001N (0.005M), LabChem™

Supplier:  LabChem LC156004

 View more versions of this product

Catalog No. LC156004

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Iodine
Red through Brown
Liquid
I2
MFCD00011355 MFCD00164163
Soluble in water
II
253.81
CHEBI:17606
Certified
Passes Test
7553-56-2,7732-18-5,7681-11-0,7647-01-0
0.13,99.6,0.25,0.02
4 L
I2
iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
PNDPGZBMCMUPRI-UHFFFAOYSA-N
diiodine
807
253.809
0.0100N ±0.0001N (0.005M)
Amber Glass

Chemical Identifiers

7553-56-2
253.81
PNDPGZBMCMUPRI-UHFFFAOYSA-N
CHEBI:17606
II
I2
MFCD00011355 MFCD00164163
807
diiodine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions