missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodine-Bromine Solution, Hanus, Spectrum™ Chemical
Specifications
Specifications
| Chemical Name or Material | Iodine-Bromine Solution |
| CAS | 7553-56-2, 7726-95-6, 64-19-7 |
| CAS Max % | 1.23, 0.87, 97.9 |
| Grade | Chemical Solution |
| Packaging | Amber Glass Bottle |
| Quantity | 500 mL |
CAUTION: For laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.