missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodine 0.2 Normal DEA List I, Reagents
Supplier: Labchem Inc CI100000500C
Alert:
DEA LIST I
Description
- Iodine 0.2 N
- DEA List I
- Appearance: Dark Brown Liquid
- Specific Gravity: 1.059
- Brand: Reagents
- NIST Traceable
- Prepared with ACS Grade Components and ASTM Type II Water
Specifications
| Liquid or Solid | |
| Blend | |
| Laboratory |
| 500 mL | |
| 0.200 N |
Chemical Identifiers
| Blend |