missing translation for 'onlineSavingsMsg'
Learn More

Iodine 0.2 Normal DEA List I, Reagents

Catalog No. p-200104727
Change view
Click to view available options
Quantity:
500 mL
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
361059168 4 L
361059145 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
DEA LIST I
Catalog No. 361059168 Supplier Labchem Inc Supplier No. CI1000004C
Only null left
DEA LIST I
  • Iodine 0.2 N
  • DEA List I
  • Appearance: Dark Brown Liquid
  • Specific Gravity: 1.059
  • Brand: Reagents
  • NIST Traceable
  • Prepared with ACS Grade Components and ASTM Type II Water

Chemical Identifiers

Molecular Weight (g/mol) Blend

Specifications

Physical Form Liquid or Solid
Quantity 4 L
Percent Purity 0.200 N
Grade Laboratory