missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Iodine, 0.1N Standardized Solution

Catalog No. p-4408553
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA35634K2 1 L
AA35634K7 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA35634K2 Supplier Thermo Scientific Chemicals Supplier No. 035634K2
Only null left

CAS: 7553-56-2 | I2 | 253.81 g/mol

Iodine, 0.1N standardized solution is used for accurate volumetric analysis. It is also used as a disinfectant and antibacterial agent in solutions, tinctures and salves.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7553-56-2
Molecular Formula I2
Molecular Weight (g/mol) 253.81
MDL Number MFCD00011355 MFCD00164163
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
PubChem CID 807
ChEBI CHEBI:17606
SMILES II

Specifications

Physical Form Liquid
Solubility Miscible with chloroform, carbon tetrachloride, carbon disulfide, cyclohexane, methanol, ethyl acetate, toluene, benzene, n-hexane, butan-2-ol, bromoethane, n-heptane, glycerol and diethyl ether
IUPAC Name molecular iodine
Quantity 1 L
Formula Weight 253.81
Concentration or Composition (by Analyte or Components) Iodine: 1.3%, Potassium iodide: 2.5%, Water: 96.2%
Chemical Name or Material Iodine
Hazard Category H335-H373
Precautionary Statement P260-P314-P501c
EINECSNumber 231-442-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only