missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Iodine, 0.1N Standardized Solution
Description
Iodine, 0.1N standardized solution is used for accurate volumetric analysis. It is also used as a disinfectant and antibacterial agent in solutions, tinctures and salves.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Physical Form | Liquid |
| Solubility | Miscible with chloroform, carbon tetrachloride, carbon disulfide, cyclohexane, methanol, ethyl acetate, toluene, benzene, n-hexane, butan-2-ol, bromoethane, n-heptane, glycerol and diethyl ether |
| Type | Analytical Volumetric Solution |
| IUPAC Name | diiodine |
| Quantity | 4 L |
| Formula Weight | 253.81 |
| Grade | Standard |
| Concentration or Composition (by Analyte or Components) | Iodine: 1.3%, Potassium iodide: 2.5%, Water: 96.2% |
| Chemical Name or Material | Iodine |
Safety and Handling
| Hazard Category | H335-H373 |
| Precautionary Statement | P260-P314-P501c |
| EINECSNumber | 231-442-4 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only