missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodine, 0.01 N (0.005 M) Solution, Spectrum™ Chemical
$107.56 - $298.24
Chemical Identifiers
| CAS | 7553-56-2 |
|---|---|
| Molecular Formula | I2 |
| Molecular Weight (g/mol) | 253.81 |
| MDL Number | MFCD00011355 MFCD00164163 |
| InChI Key | PNDPGZBMCMUPRI-UHFFFAOYSA-N |
| IUPAC Name | diiodine |
| SMILES | II |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18611046
|
Spectrum Chemical Mfg Cor
I091500MLGL |
500 mL | Amber Glass Bottle |
Each for $107.56
|
|
||||
|
18611044
|
Spectrum Chemical Mfg Cor
I0911LTGL |
1 L | Amber Glass Bottle |
Each for $128.24
|
|
||||
|
18611045
|
Spectrum Chemical Mfg Cor
I0914LTGL |
4 L | Amber Glass Bottle |
Each for $298.24
|
|
||||
Chemical Identifiers
| 7553-56-2 | |
| 253.81 | |
| PNDPGZBMCMUPRI-UHFFFAOYSA-N | |
| II |
| I2 | |
| MFCD00011355 MFCD00164163 | |
| diiodine |
Specifications
| 7553-56-2,7732-18-5,7681-11-0 | |
| I2 | |
| PNDPGZBMCMUPRI-UHFFFAOYSA-N | |
| diiodine | |
| 0.0100 ± 0.0005 N |
| 500 mL | |
| MFCD00011355 MFCD00164163 | |
| II | |
| 253.81 | |
| Amber Glass Bottle |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.