missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodine, 0.01 N (0.005 M) Solution, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor I0914LTGL
Specifications
| 7553-56-2,7732-18-5,7681-11-0 | |
| I2 | |
| PNDPGZBMCMUPRI-UHFFFAOYSA-N | |
| diiodine | |
| 0.0100 ± 0.0005 N |
| 4 L | |
| MFCD00011355 MFCD00164163 | |
| II | |
| 253.81 | |
| Amber Glass Bottle |
Chemical Identifiers
| 7553-56-2 | |
| 253.81 | |
| PNDPGZBMCMUPRI-UHFFFAOYSA-N | |
| II |
| I2 | |
| MFCD00011355 MFCD00164163 | |
| diiodine |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.