missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UltraPure™ Low Melting Point Agarose
GREENER_CHOICE

Catalog No. 16520050
Encompass
Change view
Click to view available options
Quantity:
50 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16520050 50 g
16520100 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 16520050 Supplier Invitrogen™ Supplier No. 16520050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Easy-pour spout reduces spills and contamination

UltraPureLow Melting Point Agarose is a polysaccharide used for size-based separation of nucleic acids in agarose gel electrophoresis applications. UltraPureLow Melting Point Agarose is ideal for resolving DNA and RNA fragments larger than 1,000bp and for the recovery of nucleic acid fragments after electrophoresis, as it melts at 65.5°C (below the melting point of most nucleic acids). The solution remains fluid at 37°C and will set rapidly at temperatures below 25°C

  • Ideal for analysis and recovery of DNA and RNA for routine applications
  • Strong gel structure allows for better handling and less breakage
  • Ideal for variety of applications such as in-gel manipulation, tissue and cell culture, and viral plaque assays
  • Can also be used in protein electrophoresis applications such as Ouchterlony (antigen-antibody interaction assay) and radial immunodiffusion (RID) (antigen quantitation assay)
  • Improved packaging: Environmentally friendly, sustainable packaging uses 75% less plastic than the original bottles. This means less energy and raw material used in manufacture, and less waste in Landfills
  • Performance and quality testing: Performance of UltraPure Agarose is evaluated to satisfy specifications in appearance, moisture, gel strength, gelling temperature, melting temperature, sulfate content, electroendosmosis, and DNase/RNase assay
  • Separation range: >1,000 bp.

UltraPureLow Melting Point Agarose (Cat. No. 16520-100) is a replacement for the item previously sold under Cat. No. 15517-022.

Agarose Gel Electrophoresis, DNA & RNA Purification & Analysis, Nucleic Acid Gel Electrophoresis & Blotting

Order Info

Shipping Condition: Room Temperature

Specifications

Content And Storage UltraPure™ Agarose products are packaged in new eco-friendly pouches that use less material than conventional plastic bottles. Store at 15° to 30°C.
Separation Range 100 to >30000 bp
Form Powder
For Use With (Application) Nucleic Acid Gel Electrophoresis, Protein Electrophoresis, Blotting
Gel Compatibility Agarose Gels
Melting Point Low Melting Point
Quantity 50 g
Shipping Condition Room Temperature
Product Line UltraPure
Product Type Agarose Gel
I want to pour my own gels. Which agarose should I use?

Our UltraPure Agarose is standard melting-point agarose designed for routine separation and analysis of DNA and RNA fragments in the 500-23,000 nt range. UltraPure Agarose 1000 is a specialized agarose that provides higher resolution of PCR fragments and other short DNA fragments. We also offer an UltraPure Low Melting Point Agarose, which is ideal for resolving DNA fragments from 10 to 1,000 bp with a low melting temperature of 65°C or less.

How can generation of replication competent adenoviruses be avoided when using your pSilencer adeno 1.0-CMV System?

Although there is only a very small chance of creating replication competent virus, steps should still be taken to avoid added risk. The virus is expanded in two rounds of amplification, and all secondary expansions should be performed from an initial expansion stock. Secondary amplifications in HEK293 cells should not be performed from the stock of another secondary expansion, as this could increase the chance of making replication competent virus. See the product manual for more detail and guidelines for screening.

For Research Use Only. Not for use in diagnostic procedures.