missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UltraPure™ Agarose
GREENER_CHOICE

Catalog No. 16500500
Encompass
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16500500 500 g
16500100 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 16500500 Supplier Invitrogen™ Supplier No. 16500500
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Easy-pour spout reduces spills and contamination

UltraPure™ Agarose is a polysaccharide used for size-based separation of nucleic acids in agarose gel electrophoresis applications. UltraPure™ Agarose is ideal for resolving DNA and RNA fragments from 100 bp to >30 kb

  • Ideal for analysis and recovery of DNA and RNA for routine applications
  • Strong gel structure allows for better handling and less breakage
  • Can be used in protein electrophoresis applications such as Ouchterlony (antigen-antibody interaction assay) and radial immunodiffusion (RID) (antigen quantitation assay)
  • Improved packaging
  • Performance and quality testing

Agarose Gel Electrophoresis, DNA & RNA Purification & Analysis, Nucleic Acid Gel Electrophoresis & Blotting

Order Info

Shipping Condition: Room Temperature

Specifications

Content And Storage Contents: 1 pouch containing 500 g of UltraPure™ Agarose
Storage: 15°C to 30°C
Separation Range 100 to >30000 bp
Form Powder
For Use With (Application) Nucleic Acid Gel Electrophoresis, Protein Electrophoresis, Blotting
Gel Compatibility Agarose Gels
Melting Point Standard Melting Point
No. of Reactions 2500
Quantity 500 g
Shipping Condition Room Temperature
Product Line UltraPure
Product Type Agarose Gel
Show More Show Less
I want to pour my own gels. Which agarose should I use?

Our UltraPure Agarose is standard melting-point agarose designed for routine separation and analysis of DNA and RNA fragments in the 500-23,000 nt range. UltraPure Agarose 1000 is a specialized agarose that provides higher resolution of PCR fragments and other short DNA fragments. We also offer an UltraPure Low Melting Point Agarose, which is ideal for resolving DNA fragments from 10 to 1,000 bp with a low melting temperature of 65°C or less.

How can generation of replication competent adenoviruses be avoided when using your pSilencer adeno 1.0-CMV System?

Although there is only a very small chance of creating replication competent virus, steps should still be taken to avoid added risk. The virus is expanded in two rounds of amplification, and all secondary expansions should be performed from an initial expansion stock. Secondary amplifications in HEK293 cells should not be performed from the stock of another secondary expansion, as this could increase the chance of making replication competent virus. See the product manual for more detail and guidelines for screening.

For Research Use Only. Not for use in diagnostic procedures.