Learn More
Invitrogen™ UltraPure™ SDS Solution, 10%
Filtered and ready to use
$151.00 - $234.00
Specifications
| Product Type | SDS Solution |
|---|---|
| Form | Liquid |
| Product Line | UltraPure |
| Reagent Type | SDS |
| Shipping Condition | Room Temperature |
Description
UltraPure 10% SDS Solution is a premixed aqueous solution containing 10% sodium dodecyl sulfate (SDS). SDS is a protein denaturant used in polyacrylamide gel electrophoresis for molecular weight determinations.
- Used to dissociate nucleic acid-protein complexes in DNA extraction protocols, to disrupt cell membranes, and to prepare prehybridization and hybridization solutions
- Sustainable packaging
DNA & RNA Purification & Analysis, DNA Extraction, Gene Expression Analysis & Genotyping, General gDNA Purification Reagents and Accessories, Genomic DNA Purification, Microarray Analysis, Northern Blotting, Nucleic Acid Gel Electrophoresis and Blotting, Southern Blotting
Order Info
Shipping Condition: Room Temperature
Specifications
| SDS Solution | |
| UltraPure | |
| Room Temperature |
| Liquid | |
| SDS |
For Research Use Only. Not for use in diagnostic procedures.