missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UltraPure™ SDS Solution, 10%
GREENER_CHOICE

Catalog No. 24730020
Encompass
Change view
Click to view available options
Quantity:
1 L
4 x 100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24730020 1 L
15553027 4 x 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 24730020 Supplier Invitrogen™ Supplier No. 24730020

Filtered and ready to use

UltraPure 10% SDS Solution is a premixed aqueous solution containing 10% sodium dodecyl sulfate (SDS). SDS is a protein denaturant used in polyacrylamide gel electrophoresis for molecular weight determinations.

  • Used to dissociate nucleic acid-protein complexes in DNA extraction protocols, to disrupt cell membranes, and to prepare prehybridization and hybridization solutions
  • Sustainable packaging

DNA & RNA Purification & Analysis, DNA Extraction, Gene Expression Analysis & Genotyping, General gDNA Purification Reagents and Accessories, Genomic DNA Purification, Microarray Analysis, Northern Blotting, Nucleic Acid Gel Electrophoresis and Blotting, Southern Blotting

Order Info

Shipping Condition: Room Temperature

Specifications

Content And Storage 1 bottle containing 1 L of Ultrapure SDS at 10% concentration (w/v). Store at room temperature.
Concentration 0.1
Form Liquid
For Use With (Application) Nucleic Acid Gel Electrophoresis, Blotting
Quantity 1 L
Reagent Type SDS
Shipping Condition Room Temperature
Product Line UltraPure
Product Type SDS Solution
Why do solutions containing SDS form precipitates?

SDS is a detergent used to denature proteins leading to disruption of cell membranes and dissociation of nucleic acid-protein complexes in DNA and RNA extractions. It tends to come out of solution at concentrations of 10% or greater, especially at low temperatures or in high-salt buffers. SDS can be resolubilized by slowly heating the solution to 65°C with slight agitation.

For Research Use Only. Not for use in diagnostic procedures.