missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Total RNA Lysis Solution, Nucleic Acid Purification

Catalog No. 4305895
Encompass
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4305895 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4305895 Supplier Invitrogen™ Supplier No. 4305895

Total RNA Lysis Solution is a 2X concentrate ideal for the chemical disruption of cells and tissue (cultured cells, primary cell isolates, and animal/plant tissues) prior to RNA or DNA extraction.

Total RNA Lysis Solution is a 2X concentrate ideal for the chemical disruption of cells and tissue (cultured cells, primary cell isolates, and animal/plant tissues) prior to RNA or DNA extraction. Following downstream purification (e.g., using the 6100 PrepStation), intact RNA can be isolated that is free of contaminating protein and metal ions. Total RNA Lysis solution is the recommended reagent for the Total RNA Isolation Chemistry System for the ABI PRISM™ 6100 Nucleic Acid PrepStation.

Order Info

Shipping Condition: Room temperature

Specifications

Concentration 2X
Content And Storage Store at room temperature.
For Use With (Equipment) 6100 Nucleic Acid PrepStation
No. of Reactions 100 Preps
Quantity 250 mL
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.