missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TE Buffer (20X), RNase-free

Essential to success of RiboGreen™ RNA quantitation assays

Supplier:  Invitrogen™ T11493

Encompass

Catalog No. T11493


Only null left

Description

Description

RNase-free TE Buffer (20X) is an essential component of Qubit and Quant-iT assays, including PicoGreen, RiboGreen, and Qubit dsDNA HS. It is offered here as a standalone solution to support quantification of dsDNA, ssDNA and RNA.

Note: For dilution instructions, storage considerations, and assay compatibility information please see the Product Information Sheet (Pub. No. MAN1000402) below.

Order Info

Shipping Condition: Room temperature

Specifications

Specifications

Store at room temperature.
TE Buffer
Room Temperature
20X
100 mL
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.