missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Tali™ Calibration Beads

Catalog No. T10790
Encompass
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T10790 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T10790 Supplier Invitrogen™ Supplier No. T10790

Includes

Tali - Green Calibration Beads, Tali - Red Calibration Beads, Tali - Alignment Beads

Align and calibrate Tali™ Image-Based Cytometer every three months

Flp-In 293 T-REx cell lines is designed for rapid generation of stable cell lines that ensure homogenous expression of protein of interest from a Flp-In expression vector. Targeted integration of a Flp-In expression vector ensures high-level expression of gene of interest. The Flp-In T-REx™-293 Cell Line contains pFRT/lacZeo and pcDNA™6/TR (from the T-REx™ System) stably integrated. Co-transfection of the Flp-In Cell Lines with a Flp-In expression vector and the Flp recombinase vector, pOG44, results in targeted integration of the expression vector to the same locus in every cell, ensuring homogeneous levels of gene expression.
The Flp-In-293 work well with Flp-In vectors that express a gene from the CMV promoter (e.g., pcDNA™5/FRT, pcDNA5/FRT/V5-His-TOPO™, and pSecTag/FRT/V5-His-TOPO™).

Cell Analysis, Cell Counting and Viability, Cell Counting, Viability, and Cryopreservation, Cell Culture, Cellular Imaging, Imaging Cytometry

Order Info

Shipping Condition: Room temperature

Specifications

Product Type Calibration Bead
For Use With (Equipment) Tali™ Image-Based Cytometer
Content And Storage Tali™ - Green Calibration Beads: Store 2 – 6°C
Tali™ - Red Calibration Beads: Store 2 – 6°C
Tali™ - Alignment Beads; Store 2 – 6°C
Color Red, Green
Shipping Condition Room Temperature
Format Kit
Product Line Tali
Quantity 1 kit

For Research Use Only. Not for use in diagnostic procedures.