missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SuperScript™ II Reverse Transcriptase

Catalog No. 18064071
Encompass
Change view
Click to view available options
No. of Reactions:
10 Reactions
200 Reactions
50 Reactions
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
18064071 200 Reactions
18064022 10 Reactions
18064014 50 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18064071 Supplier Invitrogen™ Supplier No. 18064071

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Invitrogen SuperScript II Reverse Transcriptase is a genetically engineered MMLV reverse transcriptase (RT) with reduced RNase H activity and increased thermal stability compared to wild-type MMLV RT.

Invitrogen SuperScript II Reverse Transcriptase is a genetically engineered MMLV reverse transcriptase (RT) with reduced RNase H activity and increased thermal stability compared to wild-type MMLV RT. Mutations in the RNase H domain of the enzyme eliminate degradation of the RNA during first-strand cDNA synthesis, which results in higher yields of full-length cDNA.

SuperScript reverse transcriptases are the most highly trusted and widely used RTs with over 50,000 citations, reviews, and publications to date. The enzyme is purified to near homogeneity from E. coli containing the modified pol gene of Moloney Murine Leukemia Virus.

Features of SuperScript II Reverse Transcriptase include:
• High cDNA yields and more full-length product
• Generation of cDNA up to 12.3 kb
• First-strand cDNA synthesis at higher temperatures than conventional MMLV RT

Note: The latest member of the SuperScript RT family, SuperScript IV Reverse Transcriptase (Cat. No. 18090010), features enhanced thermostability, processivity, yields, and performance with any RNA samples, including those of suboptimal purity or integrity.

Order Info

Shipping Condition: Wet ice

Specifications

Concentration 200 U/μL
Content And Storage

• SuperScript II Reverse Transcriptase, 4 x 50 μL (4 x 10,000 units at 200 U/μL)
• 5X First-strand buffer
• DTT (100 mM)

Store at –20°C.

Format Stand-alone Enzyme
GC-Rich PCR Performance High
Reaction Speed 50 min.
Technique Reverse Transcription
Optimal Reaction Temperature 42°C
Quantity 4 x 10,000 units
Reverse Transcriptase SuperScript II
Ribonuclease H Activity Reduced
Shipping Condition Wet Ice
Final Product Type First-Strand cDNA
No. of Reactions 200 Reactions
Reaction Format Separate components
Reagent Type Reverse Transcription
Size (Final Product) Up to 12.3 kb
Starting Material RNA
Show More Show Less
How can I remove genomic DNA contamination from my sample prior to performing RT-PCR?

We recommend using ezDNase (Cat. No. 11766051). ezDNase Enzyme's high specificity for double-stranded DNA enables efficient and fast genomic DNA removal without reduction in the quality or quantity of RNA. ezDNase Enzyme is heat-labile and so can be easily deactivated by heat treatment at moderate temperature (55 degrees C). These features make ezDNase Enzyme an excellent choice for genomic DNA removal prior to reverse transcription reactions.

How much RNA should be employed for first-strand cDNA synthesis?

The amount of RNA template for a cDNA synthesis is highly flexible and depends upon the amount of sample available and an individual's need. In general, 1 µg total RNA is used in a typical 20-µL RT reaction.

Should I treat the cDNA with RNase H prior to downstream processing?

RNase H treatment is not always necessary. Many PCR reactions work without it. However, for cDNA synthesized with RNase H-deficient reverse transcriptases (like SuperScript II, III, and IV), RNA/cDNA hybrids—especially GC-rich ones—may not denature well, reducing PCR sensitivity. RNase H treatment can help in such cases. Additionally, RNase H treatment is beneficial for cloning larger fragments.

What percentage of RNA is converted to cDNA when performing reverse transcription?

This depends highly on the quality of the sample. mRNA itself makes up 1-5% of total RNA. Depending on the primer and enzyme used, reverse transcription can covert >70% of that into cDNA.

I'm setting up my RT reaction and am trying to decide whether I should use random primers, oligo(dT) primer, gene-specific primer, or oligo(dT)/random mix primers. What would you suggest?

Random primers are the best choice for degraded RNA, RNA with heavy secondary structure, non-polyadenylated RNA, or prokaryotic RNA. It is recommended only for two-step RT-PCR, and typically gives the highest yields, although the cDNA may not necessarily be full length. Oligo(dT) primers are good to use when trying to recover full-length cDNA from 2-step RT-PCR. The reaction is influenced by secondary structure and RNA quality. Gene specific primers should be used for very specific, mainly one-step RT-PCR reactions.

The DTT in my reverse transcription kit has precipitated—can I still use it?

No, the DTT will need to be replaced.

Are SuperScript II and III RTs RNase H minus?

These enzymes contain the domains of RNase H, but they have been mutated for reduced RNase H activity. In RNase H activity detection assays, we are not able to detect any RNase H activity.

How much of the first-strand cDNA reaction should I load for PCR?

While the volume is dependent on the starting amount of RNA used for the first-strand synthesis and the abundance of the target gene, we'd recommend starting with 10% of the first-strand reaction for your PCR reaction.

What reagents inhibit SuperScript II RT activity?

The following reagents will inhibit SuperScript II RT activity by at least 50%.
34% glycerol
4 µg/mL heparin
0.0025% (w/v) SDS
5% (v/v) formamide
17.0% (v/v) DMSO
4 mg/mL glycogen
30 mM guanidine-HCl
15 mM guanidine isothiocyanate
1 mM EDTA
2.5 mM NaPPi
0.4 mM Spermidine

What is the highest temperature that MMLV, SuperScript II, SuperScript III, or SuperScript IV RTs can be used?

The optimal temperature for for MMLV is 42 degrees C. The optimal temperature for SuperScript II RT is 42 degrees C, and can be used up to 50 degrees C. The optimal temperature for SuperScript III RT is 50 degrees C, and can be used up to 55 degrees C. For some qRT-PCR reactions where gene-specific primers are used, you can do the RT reaction at 60 degrees C. The optimal temperature for SuperScript IV RT is 50 degrees C, but can be used up to 65 degrees C.

How can I tell if my RT-PCR product is RNA specific?

Include a control reaction where the RNA has not been incubated with reverse transcriptase to test for specificity. If this RNA gives a PCR product, it is most likely generated from genomic DNA contamination. Alternatively, a primer set spanning two different exons can be designed such that the PCR product from the cDNA would be of a different size compared to a product generated from genomic DNA. Primers may also be designed to span an exon/exon junctions. These primers are not likely to amplify from genomic DNA templates. For DNase treatment of RNA, we recommend using Amplification-Grade DNase I (Cat. No. 18068-015) or ezDNase Enzyme (Cat. No. 11766051).

What is the shipping condition for SuperScript II Reverse Transcriptase and SuperScript First-Strand Synthesis System for RT-PCR?

SuperScript II Reverse Transcriptase and SuperScript First-Strand Synthesis System for RT-PCR are shipped on wet ice. Our packaging and gel ice are designed to last approximately 48 hours before rising above 8 degrees C. Overnight incubation under these shipping conditions does not affect the SuperScript II Reverse Transcriptase enzyme's activity, performance, or applicable expiry date.


For Research Use Only. Not for use in diagnostic procedures.