Learn More
Invitrogen™ Spheroplast Kit for Yeast
Description
Includes
SOS medium, SE, SCE, 1M sorbitol, CaS, 40% PEG, CaT, sterile water, zymolyase, and 1M DTT. Sufficient reagents are provided in the Spheroplast Kit for 10 spheroplast preparations, each of which can be used for 5 transformations.
- Reagents have been optimized for use with Pichia pastoris and are guaranteed by manufacturer to generate 70% spheroplasts
Cloning, Pichia pastoris Expression Systems, Protein Expression, Proteins, Expression, Isolation and Analysis, Transformation, Yeast Expression
Specifications
Specifications
| Format | Liquid |
| Product Type | Spheroplast Kit |
| Content And Storage | The Spheroplast Kit contains the following reagents: SOS medium, SE, SCE, 1M sorbitol, CaS, 40% PEG, CaT, sterile water, zymolyase, and 1M DTT. Sufficient reagents are provided in the Spheroplast Kit for 10 spheroplast preparations, each of which can be used for 5 transformations. All components are guaranteed stable for 6 months when properly stored. |
| Target Organism Class | P. pastoris |
| Quantity | 1 kit |
Frequently Asked Questions (FAQs)
Generally, large colonies represent transformants containing pPIC6/pPIC6α integrants, while small colonies represent transformants containing pPIC6/pPIC6α non-integrants. These non-integrants have transduced the pPIC6/pPIC6α plasmid, and therefore, exhibit a low level of blasticidin resistance in the initial selection process. Upon subsequent screening, these non-integrant transformants do not retain blasticidin resistance.
When choosing a blasticidin-resistant transformant for your expression studies, we recommend that you pick blasticidin-resistant colonies from the initial transformation plate and streak them on a second YPD plate containing the appropriate concentration of blasticidin. Select transformants that remain blasticidin-resistant for further studies.
Here are some suggestinos:
- Make sure that you have harvested cells during log-phase growth (OD <1.0 generally).
- If electroporation is being used, see the electroporator manual for suggested conditions. Vary electroporation parameters if necessary.
- Use more DNA.
- Use freshly made competent cells.
- If the LiCl transformation method is being used, try boiling the carrier DNA.
Here are some things to consider:
- If the OD of cells that are used is too high, they will not spheroplast. Do not overgrow cells.
- Do not use old cells and make sure that they are in log phase of growth.
- Make sure to mix zymolyase well before using. Zymolyase is more of a suspension than a solution.
- Make the PEG solution fresh each time and check the pH.
Use the following high cell density protocol for pGAP clones. Feed carbon until the desired density is reached (300 to 400 g/L wet cell weight (WCW)). If the protein is well-behaved in the fermenter, increase to 300-400 g/L WCW as with methanol inducible clones. These densities can be reached in less than 48 hours of fermentation. We have fermented constitutive expressers on glycerol using these protocols with good results. Some modifications to the Fermentation Basal Salts Medium that you might want to make are:
1) Substitute 2% dextrose for the 4% glycerol in the batch medium.
2) Substitute 40% dextrose for the 50% glycerol in the fed-batch medium.
3) Feed the 40% dextrose at 12 mL/L/hr (Jim Cregg has published data on expression using several carbon sources as substrates; dextrose gave the highest levels of expression).
4) Yeast extract and peptone may be added to the medium for protein stability.
One warning: If you are working with His- strains, they remain His- after transformation with pGAPZ. Fermentation in minimal medium will require addition of histidine to the fermenter.
No, you cannot autoclave methanol. There are two approaches to this, depending a bit on the size of the bioreactor and the volumes involved. You can either dilute to working concentration and filter-sterilize with a filter suitable for alcohols, or you can just assume that methanol is sterile (it should be) and dilute into sterile water. For the ammonium hydroxide solution, you should also not autoclave it. You can assume the 30% stock solution is sterile (nothing should live in this solution) and dilute into sterile water to the working concentration.
For Research Use Only. Not for use in diagnostic procedures.