missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Safe Imager™ 2.0 Power Cord Replacement

Catalog No. G6602
Encompass
Change view
Click to view available options
For Use With (Equipment):
Safe Imager™
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
G6602 Safe Imager™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G6602 Supplier Invitrogen™ Supplier No. LSG6602
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Each instrument comes with an amber filter unit and viewing glasses

Designed for viewing stained gels on the laboratory bench top or within gel documentation systems

Light from the LED source inside the instrument passes through a blue filter producing light with a narrow emission peak centered at approximately 470nm, effective for the excitation of SYBR™ DNA-binding dyes such as SYBR Safe DNA gel stain, as well as many of our protein gel stains such as SYPRO™ Ruby, SYPRO Orange, and Pro-Q™ Diamond stains. Sensitivity obtained using this instrument is comparable to that obtained with a standard UV transilluminator. Unlike UV transilluminators, the Safe Imager transilluminator does not produce UV light and does not require UV protective equipment during use.

Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting, Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis

Specifications

Content And Storage Each instrument includes an amber filter unit and viewing glasses.
For Use With (Equipment) Safe Imager™
Type Replacement Power Cord
Quantity 1 unit
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.