missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Ribonuclease Inhibitor, Cloned

Catalog No. 15518012
Encompass
Change view
Click to view available options
Quantity:
1000 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15518012 1000 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15518012 Supplier Invitrogen™ Supplier No. 15518012

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Ribonuclease Inhibitor, Cloned is supplied at 10,000 units/mL in 20mM HEPES-KOH (pH 7.6), 50mM KCl, 8mM DTT, and 50% (v/v) glycerol.

Neutral pancreatic ribonuclease inhibitor

Ribonuclease Inhibitor, Cloned is a potent non-competitive inhibitor of neutral pancreatic ribonucleases.
Applications: Cell-free translation systems. Reverse transcription of mRNA.
Source: Recombinant, purified from E. coli.
Performance and Quality Testing: No detectable contaminating activity is observed in DNA nicking, ribonuclease, and protease assays.
Unit Definition: One unit inhibits 5ng RNase A by 50% using cytidine 2', 3' cyclic monophosphate (cCMP) as the substrate.

PCR and Real-Time PCR, Reverse Transcription

Order Info

Shipping Condition: Dry ice

Specifications

Concentration 50 mM
Content And Storage Ribonuclease Inhibitor, Cloned is supplied at 10,000 units/ml in 20 mM HEPES-KOH (pH 7.6), 50 mM KCl, 8 mM DTT, and 50% (v/v) glycerol. Store at -20°C.
Quantity 1000 U
Shipping Condition Dry Ice

For Research Use Only. Not for use in diagnostic procedures.