missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Qubit™ ssDNA Assay Kit DFS Item

Quantitate oligonucleotides or long ssDNA easily and quickly with the Qubit ssDNA Assay Kit, which works with the Qubit Fluorometer and other fluorometers or plate readers. Common contaminants such as salts, solvents, detergents, and protein are well tolerated with this assay.

Supplier:  Invitrogen™ Q10212

Catalog No. Q10212


Only null left
Explore available promotions

Requires : Qubit 1.0 or 2.0 Fluorometer or other fluorometer and 500μL thin-walled PCR tubes

Description

Description

The Qubit ssDNA Assay Kit is designed specifically for use with the Qubit Fluorometer, but may be used with any fluorometer or fluorescence plate reader. The Qubit™ ssDNA Kit is ideal for quantitating single-stranded DNA or oligonucleotides. However, it is not specific for single-stranded DNA. This assay kit will also detect double-stranded DNA and RNA but it will not detect contaminating protein or nucleotides. The assay is designed to be accurate for initial sample concentrations from 50 pg/μL to 200 ng/μL providing an assay range of 1–200 ng. The kit provides concentrated assay reagent, dilution buffer, and pre-diluted DNA standards. Simply dilute the reagent using the buffer provided, add your sample (any volume between 1 μl and 20 μl is acceptable), and read the concentration using the Qubit Fluorometer. Common contaminants, such as salts, free nucleotides, solvents, detergents, or protein are well tolerated in the assay.

Which product to choose for ssDNA quantitation?
  • For 1–20 samples: use this Qubit ssDNA Assay Kit with the Qubit Fluorometer
  • For 20–2000 samples: use the Quant-iT OliGreen ssDNA Assay Kit with microplate reader
Notes:
1. All Qubit assay kits can be used with the Qubit 1.0, Qubit 2.0, Qubit 3, and Qubit 4 fluorometers.
2. 500 μL thin-walled PCR tubes are required but not included.

Order Info

Shipping Condition: Room temperature

Specifications

Specifications

ssDNA Quantitation
500/525
0.2 to 240 ng
1 kit
Fluorescence
Qubit Fluorometer
Qubit
Room temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.