missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PureLink™ Nucleic Acid Purification Rack

Catalog No. K210013
Encompass
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K210013 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K210013 Supplier Invitrogen™ Supplier No. K210013

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

The PureLink Nucleic Acid Purification Rack consists of a: Column holder rack (for processing 12 miniprep, 8 midiprep, and 4 maniprep columns), Collection Tube Rack (capable of accommodating various types and sizes of recovery tubes), and Large capacity Waste Tray for collecting waste.

Waste reservoir nests into collection tube rack for compact storage.

  • Convenient large capacity fits multiple-sized column
  • Compact nesting feature is convenient for storage
  • PureLink Nucleic Acid Purification Rack is designed for use with columns in the PureLink HiPure Plasmid Purification Systems
  • Rack includes both Collection Tube Rack for holding various sized recovery tubes and a large-capacity Waste Reservoir
  • One rack holds as many as 12 miniprep columns, 8 midiprep columns, and 4 maxiprep columns

Order Info

Shipping Condition: Room Temperature

Specifications

Product Type Nucleic Acid Purification Rack
Format Rack
Content And Storage The PureLink™ Nucleic Acid Purification Rack includes:
• Column Holder Rack (for processing 12 miniprep, 8 midiprep, and 4 maxiprep columns)
• Collection Tube Rack (capable of accommodating various types and sizes of recovery tubes)
• Large Capacity Waste Tray (for collecting waste)
Product Line PureLink
Quantity 1 Ea.
Shipping Condition Room Temperature
How do I eliminate the RNA contamination I get in my minipreps?

Low copy number plasmids may be contaminated with RNA due to the large number of cells being processed. Increase the concentration of RNAse in the buffers 4 fold.

For Research Use Only. Not for use in diagnostic procedures.