missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human IL-8 ProQuantum Immunoassay Kit DFS Item

ELISA

$1020.00 - $4270.00

Specifications

Accession Number P10145
Assay Range 0.0128 to 5000 pg/mL
Assay Sensitivity <0.01 pg/mL
Conjugate FAM™
Detection Method Taqman™ Real-time PCR
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A35575 DFS Item
View Documents
Invitrogen™
A35575
96 Tests
Each for $1,020.00
Only null left
 
A355755 DFS Item
View Documents
Invitrogen™
A355755
5 x 96 Tests
Pack of 5 Each for $4,270.00
Only null left
 
Description

Description

Interleukin 8 (IL-8, CXCL8) is a 72 amino acid pro-inflammatory factor which belongs to the CXC subfamily of chemokines, and are bound by the cell surface receptors IL-8RA and IL-8RB. IL-8 functions as a chemoattractant and potent angiogenic factor. The expression and secretion of IL-8 can be induced by diverse inflammatory stimuli in many cells, including macrophages and endothelial cells. In endothelial cells, IL-8 is present in storage vesicles called Weibel-Palade bodies. IL-8, first isolated from osteosarcoma cells, contains the ELR-motif (N-terminal Glu-Leu-Arg amino acid sequence) and signals through the CXCR1 and CXCR2 receptors. Previous nomenclature for IL-8 includes neutrophil activating protein 1 (NAP-1), granulocyte chemotactic protein 1 (GCP-1), monocyte-derived neutrophil-activating peptide (MONAP) and protein 3-10C. IL-8 and ten other members of the CXC chemokine gene family form a chemokine gene cluster in a region mapped to chromosome 4q. Cancer studies have demonstrated a role for IL-8 in the angiogenesis and growth of tumours, and IL-8 is believed to play a role in the pathogenesis of bronchiolitis, a common respiratory tract disease caused by viral infection.
Specifications

Specifications

P10145
<0.01 pg/mL
Taqman™ Real-time PCR
Plasma, Serum, Supernatant
Cytokine Protein Quantitation
3576
Assay Dilution Buffer, Antibody-Conjugate A, Antibody-Conjugate B, Antibody-Conjugate Dilution Buffer, Standard, Master Mix, Ligase
RUO
-20°C
30 mins
0.0128 to 5000 pg/mL
FAM™
ProQuantum
Human
qPCR Instruments
GCP-1; GCP1; IL8; LECT; LUCT; LYNAP; MDNCF; MONAP; NAF; NAP-1; NAP1
FAM™
Plasma 2-5 μL, Serum 2-5 μL, Supernatant 2-5 μL
IL-8
2 hrs
Videos
SDS
Documents

Documents