missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human IL-6 ProQuantum Immunoassay Kit

ELISA
Supplier: Invitrogen™ A35573
Description
IL-6 is a gene that encodes a cytokine that plays a role in inflammation and the maturation of B cells. The protein produced by this gene is an endogenous pyrogen that can induce fever in people with autoimmune diseases or infections. The gene is primarily expressed at sites of acute and chronic inflammation, where it is secreted into the serum and induces a transcriptional inflammatory response through interleukin 6 receptor, alpha. The functioning of this gene is implicated in a wide variety of inflammation-associated disease states, including suspectibility to diabetes mellitus and systemic juvenile rheumatoid arthritis. Elevated levels of the encoded protein have been found in virus infections, including COVID-19 (disease caused by SARS-CoV-2). Diseases associated with IL6 include Kaposi Sarcoma and Rheumatoid Arthritis, Systemic Juvenile.Chemical Identifiers
| P05231 | |
| <0.05 pg/mL | |
| Taqman™ Real-time PCR | |
| Plasma, Serum, Supernatant | |
| Cytokine Protein Quantitation | |
| 3569 | |
| Assay Dilution Buffer, Antibody-Conjugate A, Antibody-Conjugate B, Antibody-Conjugate Dilution Buffer, Standard, Master Mix, Ligase | |
| RUO | |
| -20°C | |
| 30 mins |
| 0.064 to 10000 pg/mL | |
| FAM™ | |
| ProQuantum | |
| Human | |
| qPCR Instruments | |
| BSF-2; BSF2; CDF; HGF; HSF; IFN-beta-2; IFNB2; IL-6 | |
| FAM™ | |
| Plasma 2-5 μL, Serum 2-5 μL, Supernatant 2-5 μL | |
| IL-6 | |
| 2 hrs |
Specifications
| P05231 | |
| <0.05 pg/mL | |
| Taqman™ Real-time PCR | |
| Plasma, Serum, Supernatant | |
| Cytokine Protein Quantitation | |
| 3569 | |
| Assay Dilution Buffer, Antibody-Conjugate A, Antibody-Conjugate B, Antibody-Conjugate Dilution Buffer, Standard, Master Mix, Ligase | |
| 96 Tests | |
| Plasma 2-5 μL, Serum 2-5 μL, Supernatant 2-5 μL | |
| IL-6 | |
| 2 hrs |
| 0.064 to 10000 pg/mL | |
| FAM™ | |
| ProQuantum | |
| Human | |
| qPCR Instruments | |
| BSF-2; BSF2; CDF; HGF; HSF; IFN-beta-2; IFNB2; IL-6 | |
| FAM™ | |
| RUO | |
| -20°C | |
| 30 mins |