missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ pFastBac™ Dual Expression Vector

Catalog No. 10712024
Encompass
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10712024 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 10712024 Supplier Invitrogen™ Supplier No. 10712024

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

pFastBac Dual (10μg) and pFastBac Dual Control vector (4ng) are supplied lyophilized.

Features two promoters in single vector for expression of two proteins simultaneously in insect cells when using Bac-to-Bac™ Baculovirus Expression System

  • Vector has two strong promoters, polyhedrin promoter and p10 promoter, for high-level expression
  • Expression control is included that expresses both CAT and β-glucuronidase (gus)

Baculovirus Production and Titering, Insect Expression, Protein Expression, Proteins, Expression, Isolation and Analysis

Specifications

Promoter p10, Polyhedrin
Product Type Expression Vector
Content And Storage pFastBac™ Dual (10 μg) and pFastBac™ Dual Control vector (4 ng) are supplied lyophilized.

Store at -20°C. Vectors are guaranteed stable for 6 months when properly stored.
Protein Tag Untagged
Cloning Method Restriction Enzyme/MCS
Quantity 10 μg
Vector pFastBac
Product Line pFastBac
I cannot grow this white colony in liquid culture. What should I do?

The concentration of gentamicin might be too high. Try lowering the amount to 5 µg/mL and try adding more of the colony to the culture medium.

What has happened when I see blue colonies? How about colonies which are blue in the center and white on the edges?

In the case of a blue colony, the E. coli has the bacmid and the plasmid in it, allowing the cells to survive the selection process. However, because the transposition has not occurred, the LacZ gene is not disrupted. For bulls-eye colonies, this indicates that the transposition took place when the colony was growing. Re-streaking for an isolated clone from the white portion of the mixed colony should yield some colonies where transposition occurred.

I'm getting mostly white/wild-type plaques instead of blue/recombinant plaques. What am I doing wrong?

This is typically an indication of poor homologous recombination. Check the plasmid/linear DNA ratio you used. If there are some blue plaques, however, expand those viruses and check for their protein. In our experience, they are correct, even if they were in relatively low abundance.

I've infected my cells and see large polyhedra in one cell and smaller polyhedra (more numerous) in a neighboring cell. Is this normal?

Yes, cells are infected with wild-type virus individually and will develop polyhedra at different rates until all the cells in the flask are infected. The polyhedra in cells will form in approximately 3-4 days, differing in size and number until they reach their maximum capacity and burst the cell, releasing tiny particles of virus into the medium.

I'm worried that I am not getting plaques. How many days does it take to see plaques and what size are they typically?

Normally, very small white dots show up about 5-7 days and 1 mm plaques show up around day 10. Plaques can vary in size from 1 mm to 4 mm.

I forgot to add Bluo-gal to my plates. Can I add it later?

On the day you intend to pick plaques, make a solution of Bluo-gal in DMSO at 20 mg/mL. Add 50 µL per plate and spread with a glass spreader under sterile conditions. Wait 30-60 min, and your plaques should turn blue.

What might cause plates to turn blue along with blue plaques?

There are a few things that can turn plates blue:

- Too much virus when plating. Try a higher dilution.
- Cells are being singed when plated with hot melted agarose. This lyses the cells and releases lacZ into the agarose, turning it blue. Double-check plating temperatures. If plates are too wet, the blue can diffuse.

After addition of the agarose overlay to my cells, they look like crescents or are granular. What happened?

The agarose overlay was too hot. After addition of the agarose overlay, cells should still be round and healthy.

My agarose overlay is floating. Is this because I did not aspirate the medium completely?

Yes, this is indicative of an aspirating problem on the plaques. The agarose overlays were “floating” because the medium was not completely aspirated from the plates. The plates need to be completely dry before the agarose is placed over the cells, especially when plaques will be picked. To do this, we typically tip the plate slightly and keep going around the rim of the plate with the Pasteur pipette tip, being careful not to disturb the cell monolayer. If any medium pooling at the rims of the plates (they will be small pools) is seen, continue to aspirate. This “floating” agarose overlay problem may also result in wild-type contamination. The wild-type virus is able to migrate to other portions of the plates and contaminate recombinant plaques. Wild-type virus replicates much faster than recombinant virus, and can quickly overwhelm the recombinant virus.

I see what looks like small plaques, but they are too difficult to visualize. What could be the problem?

Too many cells were seeded; we recommend seeding 8 x 10e5 cells per well for a 6-well plate.

I think I've added too much virus. How important is the MOI? What happens if too much virus is used?

An MOI of 5-10 is typically used. If too much virus is added, unfortunately the cells die too soon and the protein expression level goes down.

I'm yielding no plaques from my baculovirus plaque assay. What are the possible causes for this?

The kinetics of infection may be slower than expected. Observe plates until the 8-9th day after infection. If no plaques appear, investigate the following:

- If the cells are not healthy, then poor-quality or no plaques can result. Ideally, cells should be in mid-log phase and have a viability of greater than 90%. Cells should double at least once before infection stops growth. Ensure that the correct amount of cells was used at ~70% confluency.
- The viral replication cycle can be inhibited due to poor nutritional and physical conditions of the cell.
- The temperature of the agarose is also crucial. After overlaying the agarose, the plates should be left untouched for 1 hour for the agarose to completely solidify.
- Excessive condensation during incubation at 27 degrees C can inhibit plaque formation-remove paper towels or open the container containing plates as soon as condensation appears.
- The viral titer is too low: Use a higher viral titer. You may need to re-infect your cells and collect a higher titer of your viral stock.

I am trying to clone my insert into the pFastBac vector and am getting no or few colonies after transformation. What are the possible causes and recommendations for this?

Please review the possible causes and recommendations below:

- Low transformation efficiency of competent E. coli: If stored incorrectly, prepare or obtain new competent cells. We recommend using our One Shot TOP10 or One Shot MAX Efficiency DH10B-T1R chemically competent E. coli cells.
- Impurities in DNA: Purify the insert DNA. Ensure that any excess phenol, proteins, detergents, and ethanol from the DNA solution are removed.
- Too much DNA was transformed: For chemically competent cells, add 1-10 ng of DNA in a volume of 5 µL or less per 100 µL of cells. For electrocompetent cells, add 10-50 ng of DNA in a volume of 1 µL or less per 20 µL of cells.
- Incomplete ligation reaction: Optimize your ligation reaction and include a ligation control (i.e., digested pFastBac vector + ligase; no insert).
- Ligation reaction mix inhibits transformation of competent cells: Reduce the amount of ligation reaction transformed. Dilute ligation reaction 5x with TE buffer prior to transformation.
- Problem with antibiotic: Confirm that you are using the correct antibiotic and concentration, as well as the age of the antibiotic.
- Competent cells stored improperly or handled improperly: Store competent cells at -80 degrees C; thaw cells on ice and use immediately; do not vortex.

My recombinant pFastBac construct lacks an insert. What are the possible causes and solutions for this?

Please review the possible reasons and recommendations below:

- Incomplete digestion of pFastBac plasmid or insert DNA: Use additional restriction enzyme for digestion or purify the insert DNA.
- Incomplete or excessive phosphatase treatment of pFastBac plasmid: Optimize dephosphorylation conditions according to the manufacturer's recommendations for the phosphatase you are using.
- Poor recovery of pFastBac plasmid or insert DNA from agarose gel: Use our PureLink Quick Gel Extraction System to purify high-quality plasmid DNA from your agarose gel.
- Incomplete ligation reactions: Follow ligation conditions and optimize the reaction by varying vector:insert molar ratios (1:3, 1:1, 3:1).
- Insert contains unstable DNA sequences such as LTR sequences and inverted repeats: Grow transformed cells at a lower temperature (30 degrees C) or try a different competent cell strain such as MAX Efficiency Stbl2 Competent Cells.

Can an old low-titer stock be used to make a high-titer stock?

If this lower-titer stock is a P1 or P2 stock, a viral amplification protocol can be used. If the low-titer stock was once a high-titer stock, but has dropped titer due to age or the stock was propagated many generations, then it may be necessary to regenerate the high-titer stock. If the high titer stock is >P5, then there may be an excessive amount of defective interfering particles that infect cells but do not properly replicate or produce protein. If the existing stock is plated out and a fresh plaque is re-isolated (DIPs do not form plaques), a new high-titer stock can be established.

Can a P3 viral stock be used to generate more viruses? How about a P4 or P5 stock?

Yes, the same protocol used to make your P2 viral stock can be used to make a P3, P4, or P5 viral stock. We don't recommend making the stock higher than P5, as more defective interfering particles will be produced and a decrease in protein expression level will occur.

Can the bacluovirus infect mammalian cells? How about Drosophila cells?

Yes, baculovirus can infect mammalian cells, although only at very high titers. Baculovirus works best in liver cells. However, there is no danger of cross-contamination unless the cells are directly infected with the high-titer stocks. Bacuolvirus can infect Drosophila cells; however, it will not replicate in these cells. The promoters used to drive expression of your gene in a typical baculovirus system are both late promoters and require earlier proteins from the baculovirus genome. Thus, they will not work in S2 cells since the early proteins are not made.

How many cells need to be infected in a 12-well plate with 1 plaque?

Typically, 0.5 x 106 cells per well in 2.5-3 mL is a good starting point. Lysis should begin by day 3. Virus may be harvested and amplified between 3 and 7 days (90% cell death).

I'd like to perform PCR on a plaque, and also make a stock from the same plaque. How do you suggest I do this?

Our R&D team will typically pick a plug and add it to a 12-well dish with 0.5 x 10e6 cells/well and 2.5 mL total volume per well. After approximately 3 days, remove 0.75 mL to make DNA for PCR and keep the remaining medium in an Eppendorf tube as your P1 viral stock. As an aside, it is okay to pick a plaque and store it in Grace's medium.

What is the optimal time for harvesting high-titer virus following infection? What happens if I let the cells go longer?

We recommend harvesting high-titer virus when there is 90% cell lysis. This takes approximately 5-7 days. If the cells go longer, the proteases released from the lysed cells will start to degrade viral surface proteins and result in less infectious virus.

After infecting T25 or T75 flasks with virus, when do you begin to see cell lysis and what percentage of cells will be lysed at certain time points?

This is dependent on how much virus is added. If cells are infected at an MOI of 5, usually cells are infected at 24 hours, and cells begin to lyse at around 65 hours. If less virus is used, this takes longer, and more virus takes less time.

To propagate more recombinant virus stock, what MOI should I use and when should I harvest the virus?

When propagating virus stock, use a low MOI (0.03-0.1) in order to avoid effects of defective interfering particles (DIPs). A low MOI, which ensures no more than 1 virion per cell, prevents the amplification of DIPs. A harvest time based on 15% cell viability is appropriate. NOTE: DIPs are nearly normal virus capsids containing genomes that are defective and are unable to undergo successful replication. While this "particle" is not infectious by itself, it can replicate when co-infected with normal virion, or with some other types of DI particles.

Is there a way to make baculovirus plaques more visible or distinctive?

You can stain the monolayer with neutral red or MTT to make the plaques more visible. Alternatively, you can allow the plates to develop for a few days longer (2-5 days on average) at room temperature to increase the contrast in recombinant plaques. However, the plaques stained with neutral red cannot be used for plaque purification and viral amplification.

What viral titer do you suggest having to infect my cells?

We suggest using a viral stock with a titer of >1 x 10e8 pfu/mL for expression studies.

What is the equation to calculate viral titer? Do you have an example I can use?

Please see the equation below:
pfu/mL = number of plaques (pfu)/dilution factor x mL of inocula
So, if you have a well with viral dilution of 10-8 containing 18 white plaques, the viral titer is calculated as followed:
X pfu/mL = 18 pfu/10-8 x 1 mL
X = 1.8 x 10e9 pfu/mL

Can you outline the main steps of performing a plaque assay, and any suggestions when performing this assay?

Please see the method below for an outline of the main steps of performing a plaque assay:

- Plate cells at 80% confluency in a 6-well plate
- Make a serial dilution of the P1 viral stock (1-10-5) and add to cells
- Incubate for an hour at 27 degrees C
- Mix 1% melted agarose into the medium
- Remove the viral supernatant
- Overlay the cells with the medium containing agarose
- Leave the plates for 2-3 hours for agar to completely solidify
- Incubate plates for 10-14 days
- Count plaques

When performing this assay, we suggest:

- Use cells that are in excellent health, of low passage (10-20) in log-phase growth, and high viability (>95%)
- Check viral stock for sterility (free of contamination)
- Use high-quality, low melting point agarose
- The temperature of the medium with agarose is crucial-too hot, cells will die; but if too cold, it will solidify too quickly
- Wait 2-4 hours before removing the plate after overlay so that the agarose can 100% solidify
- Count plaques on a dilution plate where (1/dilution) x # of plaques = pfu/mL
e.g., if you have 50 plaques on the 10-6 plate, then you have 1(10-6) x 50 = 5 x 10e7 pfu/mL

How do I determine the titer of my viral stock?

We recommend you perform a plaque assay to determine the titer of your viral stock. You may also perform a plaque assay to purify a single viral clone, if desired.

I performed my PCR and see different banding patterns. What do the different PCR sizes mean?

The expected PCR size will depend on the pFastBac vector that got transposed into the bacmid:

- Bacmid alone: 300 bp
- With pFastBac 1: 2300 bp + size of insert
- With pFastBac 1-Gus: 4200 bp
- With pFastBac HT: 2430 bp + size of insert
- With pFastBac HT-CAT: 3075 bp
- With pFastBac Dual: 2560 bp + size of insert
- With pFastBac Dual-Gus/CAT: 5340 bp
- With pDEST8: 2316 bp + size of insert
- With pDEST10: 2484 bp + size of insert
- With pDEST20: 3031 bp + size of insert

What are the sequencing primers for pFastBac vectors?

5' sequencing primer: 5' - TAT TCC GGA TTA TTC ATA CC - 3'
3' sequencing primer: 5' - TTC AGG TTC AGG GGG AGG TG - 3'

Do I need to include a Kozak sequence for expression of recombinant proteins in insect cells?

While the importance of a Kozak consensus sequence in translation initiation has been demonstrated in mammalian cells, there seems to be some debate as to whether the Kozak rules are as stringent in insect cells. The only way to determine its importance would be a direct comparison of expression of the same protein from different initiation sequences. Even then, the rules for optimal expression of one protein may not hold for another. Here are two references which indicate that a Kozak consensus sequence does not have any effect on efficiency of expression in insect cells:

- Hills D, Crane-Robinson C (1995) Baculovirus expression of human basic fibroblast growth factor from a synthetic gene: role of the Kozak consensus and comparison with bacterial expression.
- Biochim Biophys Acta 1260(1):14-20.
- Ranjan A, Hasnain SE (1995) Influence of codon usage and translational initiation codon context in the AcNPV-based expression system: computer analysis using homologous and heterologous genes. Virus Genes 9(2):149-153.

Is it possible to co-infect insect cells with two different recombinant viruses in order to co-express, for example, two subunits of a protein?

Yes, it is possible. Several five-subunit proteins, such as human replication factor C, have been expressed using recombinant baculovirus. We recommend that a separate high-titer stock (HTS) of each subunit be produced to optimally express the multi-subunit protein. This way, the amount of each subunit expressed can be controlled by varying the multiplicity of infection (MOI) of each subunit's HTS. Please refer to the following articles for more information:

- Chen W and Bhal OP (1991) Recombinant carbohydrate and selnomethionyl variants of human choriogonadotropin. J Biol Chem 266(13):8192-8197.
- Chen WY and Bhal OP (1991) Selenomethionyl analog of recombinant human choriogonadotropin. J Biol Chem 266(15):9355-9358.
- Fabian JR, Kimball SR, Jefferson LS (1998) Reconstitution and purification of eukaryotic initiation factor 2B (eIF2B) expressed in Sf21 insect cells. Protein Expr Purif 13(1):16-22.

What is the multiplicity of infection, and how can I calculate it?

The MOI, or multiplicity of infection, is the average number of viral particles that infect a single cell in a specific experiment. You can calculate the MOI with the following equation:
MOI (pfu/cell) = [titer (pfu) x viral stock volume (mL) used in inocula] / [cell density (cells/mL) x culture volume (mL)]

Can I scale-up the production of recombinant protein using the baculovirus expression system? If so, what methods are available?

Yes, large-scale expression experiments can be performed. Please see below for different large-scale methods, requirements, added benefits, and references:
- Stirred bioreactor
- Airlift fermentor
- Insect larvae

How can I store my viral stock?

If the medium is serum-free, add serum to 10%. Serum proteins act as substrates for proteases and therefore prevent degradation of viral coat proteins. Store viral stocks at 4 degrees C, and protect from light. Aliquots can be stored at -80 degrees C, but viral titer should be checked before use, as freeze/thaw cycles of the virus can result in a 10- to 100-fold decrease in viral titer.

Do I need to purify my recombinant virus away from an uncut or non-recombinant viral DNA?

Yes. Contamination of your recombinant DNA with uncut occlusion body positive (occ+) DNA will lead to dilution of your recombinant virus over time because, in general, uncut (wild-type, occ+) virus infects and replicates at higher efficiency than recombinant virus. Also, initiating expression studies with a pure, single virus population will ensure reproducible results.

What does viral infection look like in early, late, and very late stages?

Please see the description below of the different stages of viral infection:

Early
- Increased cell diameter-a 25-50% increase in the diameter of the cells may be observed.
- Increased size of cell nuclei-the nuclei may appear to "fill" the cells.

Late
- Cessation of cell growth-cells appear to stop growing when compared to a cell-only control.
- Granular appearance
- Signs of viral budding-vesicular appearance of cells.
- Viral occlusions-few cells will contain occlusion bodies, which appear as refractive crystals in the nucleus of the insect cell.
- Detachment-cells release from the dish or flask.

Very late
- Cell lysis-a few cells may fill with occluded virus, die, and burst, leaving signs of clearing in the monolayer.

What should I look for to indicate a successful transfection in which baculovirus is produced?

Adherent Sf9 cells round up and show a smaller contact point. Infected Sf9 cells in suspension culture round up and look larger when infected.

What are some general suggestions for transfection in baculovirus expression systems?

Please follow the recommendations below:

- Cells should be in excellent health, of their low passages (5-15), in log-phase growth, with viability >95%
- DNA must be of high purity, free of endotoxin
- No antibiotics should be used during transfection
- Cellfectin reagent has to be completely resuspended
- Include controls (media control, DNA control, and transfection reagent control) for comparison and troubleshooting

When should I harvest my protein after I have inoculated my insect cells with recombinant virus?

Peak expression of protein in insect cells is dependent on the multiplicity of infection (MOI), expression time, and the protein being expressed. Guidelines to optimize your system include using an MOI of 5-10 and an expression time of 48-72 hours. Protein expressed at times later than 72 hours may be processed aberrantly, because the large virus load can cause a breakdown of cellular processes.

Is baculovirus good for expressing toxic proteins?

Yes, baculovirus is a good candidate for the problem of expressing toxic proteins (i.e., membrane proteins). The polyhedron promoter does not express at maximal levels until 18-24 hr after infection. The polyhedron promoter is active late in the lytic cycle. That being said, it is minimally active as early as 8 hours, so if the gene is very toxic, there may be a problem. The solution in that case would be to switch to an inducible expression system. Transmembrane proteins can often be difficult to express in any system.

Do I need to include a ribosomal binding site (RBS/Shine Dalgarno sequence) or Kozak sequence when I clone my gene of interest?

ATG is often sufficient for efficient translation initiation although it depends upon the gene of interest. The best advice is to keep the native start site found in the cDNA unless one knows that it is not functionally ideal. If concerned about expression, it is advisable to test two constructs, one with the native start site and the other with a Shine Dalgarno sequence/RBS or consensus Kozak sequence (ACCAUGG), as the case may be. In general, all expression vectors that have an N-terminal fusion will already have a RBS or initiation site for translation.

Can you tell me the difference between a Shine-Dalgarno sequence and a Kozak sequence?

Prokaryotic mRNAs contain a Shine-Dalgarno sequence, also known as a ribosome binding site (RBS), which is composed of the polypurine sequence AGGAGG located just 5’ of the AUG initiation codon. This sequence allows the message to bind efficiently to the ribosome due to its complementarity with the 3’-end of the 16S rRNA. Similarly, eukaryotic (and specifically mammalian) mRNA also contains sequence information important for efficient translation. However, this sequence, termed a Kozak sequence, is not a true ribosome binding site, but rather a translation initiation enhancer. The Kozak consensus sequence is ACCAUGG, where AUG is the initiation codon. A purine (A/G) in position -3 has a dominant effect; with a pyrimidine (C/T) in position -3, translation becomes more sensitive to changes in positions -1, -2, and +4. Expression levels can be reduced up to 95% when the -3 position is changed from a purine to pyrimidine. The +4 position has less influence on expression levels where approximately 50% reduction is seen. See the following references:

- Kozak, M. (1986) Point mutations define a sequence flanking the AUG initiator codon that modulates translation by eukaryotic ribosomes. Cell 44, 283-292.
- Kozak, M. (1987) At least six nucleotides preceding the AUG initiator codon enhance translation in mammalian cells. J. Mol. Biol. 196, 947-950.
- Kozak, M. (1987) An analysis of 5´-noncoding sequences from 699 vertebrate messenger RNAs. Nucleic Acids Res. 15, 8125-8148.
- Kozak, M. (1989) The scanning model for translation: An update. J. Cell Biol. 108, 229-241.
- Kozak, M. (1990) Evaluation of the fidelity of initiation of translation in reticulocyte lysates from commercial sources. Nucleic Acids Res. 18, 2828.

Note: The optimal Kozak sequence for Drosophila differs slightly, and yeast do not follow this rule at all. See the following references:

- Romanos, M.A., Scorer, C.A., Clare, J.J. (1992) Foreign gene expression in yeast: a review. Yeast 8, 423-488.
- Cavaneer, D.R. (1987) Comparison of the consensus sequence flanking translational start sites in Drosophila and vertebrates. Nucleic Acids Res. 15, 1353-1361.

Which strand is produced using M13K07 to make ssDNA from pPROEX HT or pFastBAC vectors?

You will get the antisense strand with pProEXHT.
You will get the + or sense strand with pFastBac I of pFastBac HT.


For Research Use Only. Not for use in diagnostic procedures.