missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ pcDNA™6/TR vector Mammalian Expression Vector

Catalog No. V102520
Encompass
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V102520 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. V102520 Supplier Invitrogen™ Supplier No. V102520

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

20μg of supercoiled vector (40μL at 0.5μg/μL) supplied in TE Buffer, pH 8.0.

Uses the complete CMV promoter and adds control elements from the bacterial tetracycline resistance operon to effectively repress and derepress transcription from this strong mammalian promoter sequences

  • Regulatory vector, pcDNA6/TR, is provided for high-level expression of tetracycline repressor (TR) protein
  • This vector expresses Blasticidin resistance gene for rapid selection of mammalian cell lines that stably express TR protein

Mammalian Expression, Protein Expression, Proteins, Expression, Isolation and Analysis, Regulated Expression, Targeted Integration

Specifications

Constitutive or Inducible System Inducible
Delivery Type Transfection
Promoter CMV
Product Type Mammalian Expression Vector
Selection Agent (Eukaryotic) Blasticidin
Content And Storage 20 μg of supercoiled vector (40 μL at 0.5 μg/μL) supplied in TE Buffer, pH 8.0. Store at -20°C.
Shipping Condition Room Temperature
Protein Tag Untagged
Quantity 20 μg
Vector pcDNA
Product Line T-REx, pcDNA
For Use With (Application) Regulated Expression
Show More Show Less
Can I use any Gateway entry vector to generate entry clones for use in RNAi applications?

No, you should use an entry vector that contains the elements necessary for RNA Polymerase III-dependent expression of your shRNA (i.e., Pol III promoter and terminator).

What is a dose response curve or kill curve? And can you outline the steps involved?

A dose response curve or kill curve is a simple method for determining the optimal antibiotic concentration to use when establishing a stable cell line. Untransfected cells are grown in a medium containing antibiotic at varying concentrations in order to determine the lowest amount of antibiotic needed to achieve complete cell death. The basic steps for performing a dose response curve or kill curve are as follows:

- Plate untransfected cells at 25% confluence, and grow them in a medium containing increasing concentrations of the antibiotic. For some antibiotics, you will need to calculate the amount of active drug to control for lot variation.
- Replenish the selective medium every 3-4 days. After 10-12 days, examine the dishes for viable cells. The cells may divide once or twice in the selective medium before cell death begins to occur.
- Look for the minimum concentration of antibiotic that resulted in complete cell death. This is the optimal antibiotic concentration to use for stable selection.

Can I create stable cell lines using pENTR/U6 entry vector or the pENTR/H1/TO vector?

Unfortunately, the pENTR/U6 vector does not contain a selection marker; therefore, only transient RNAi analysis may be performed. If you wish to generate stable cell lines, perform an LR reaction into an appropriate Gateway destination vector to generate expression clones.
The pENTR/H1/TO vector contains the Zeocin resistance gene to facilitate generation of cell lines that inducbily express the shRNA of interest. Perform a kill curve to determine the minimum concentration of Zeocin that is required to kill your untransfected mammalian cell line. Please note that Zeocin-sensitive cells do not round up and detach from the plate, but rather may increase in size, show abnormal cell shape, display presence of large empty vesicles in the cytoplasm, or show breakdown of plasma/nuclear membranes.

What loop sequence should I use when designing my shRNA for cloning? Do you have any guidelines I should follow?

You can use a loop sequence of any length ranging from 4 to 11 nucleotides, although short loops (i.e., 4-7 nucleotides) are generally preferred. Avoid using a loop sequence containing thymidines (Ts), as they may cause early termination. This is particularly true if the target sequence itself ends in one or more T nucleotides. Here are some loop sequences we recommend:

- 5' - CGAA - 3'
- 5' - AACG - 3'
- 5' - GAGA - 3'

What considerations regarding transcription initiation should I take when designing my shRNA for cloning?

Transcription of the shRNA initiates at the first base following the end of the U6 promoter sequence. In the top-strand oligo, the transcription initiation site corresponds to the first nucleotide following the 4 bp CACC sequence added to permit directional cloning. We recommend initiating the shRNA sequence at a guanosine (G) because transcription of the native U6 snRNA initiates at a G. Note the following:

- If G is part of the target sequence, then incorporate the G into the stem sequence in the top-strand oligo and add a complementary C to the 3' end of the top-strand oligo.
- If G is not the first base of the target sequence, we recommend adding a G to the 5' end of the top-strand oligo directly following the CACC overhang sequence. In this case, do not add the complementary C to the 3' end of the top-strand oligo. Note: We have found that adding the complementary C in this situation can result in reduced activity of the shRNA. Alternative, if use of a G to initiate transcription is not desired, use an adenosine (A) rather than C or T. Note, however, that use of any nucleotide other than G may affect initiation efficiency and position.

How do I order the shRNA for vector expression?

Please follow the steps outlined below:

- Visit RNAi Designer
- Enter an accession number or provide a nucleotide sequence
- Determine the region for target design: ORF, 5' UTR, or 3' UTR
- Choose database for Blast
- Choose minimum and maximum G/C percentage
Select vector and strand orientation and click “RNAi Design” to design shRNA.

What molar ratio do you recommend for ligating my ds oligo to the pENTR/U6 entry vector or pENTR/H1/TO vector?

For optimal results, use a 10:1 molar ratio of ds oligo insert:vector for ligation.

How can I check the integrity of my ds oligo once it is annealed?

We suggest running an aliquot of the annealed ds oligo (5 µL of the 500 nM stock) and comparing it to an aliquot of each starting single-stranded oligo (dilute the 200 µM stock 400-fold to 500 nM; use 5 µL for gel analysis). Be sure to include an appropriate molecular weight standard. We generally use the following gel and molecular weight standard:

- Agarose gel: 4% E-Gel (Cat. No. G5000-04)
- Molecular weight standard: 10 bp DNA Ladder (Cat. No. 10821-015)

When analyzing an aliquot of the annealed ds oligo reaction by agarose gel electrophoresis, we generally see the following:
- A detectable higher molecular weight band representing annealed ds oligo.
- A detectable lower molecular weight band representing unannealed single-stranded oligos. Note that this band is detected since a significant amount of the single-stranded oligo remains unannealed.

How do I anneal my single-stranded DNA oligos to create a ds oligo?

You will want to anneal equal amounts of the top- and bottom-strand oligos to generate the ds oligos. If your single-stranded oligos are supplied lyophilized, resuspend them in water or TE buffer to a final concentration of 200 µM before use. We generally perform the annealing reaction at a final single-stranded oligo concentration of 50 µM. Annealing at concentrations lower than 50 µM can significantly reduce the efficiency. Note that the annealing step is not 100% efficient; approximately half of the single-stranded oligos remain unannealed even at a concentration of 50 µM. Please see the steps below:

1. In a 0.5 mL sterile microcentrifuge tube, set up the following annealing reaction at room temperature.
“Top-strand” DNA oligo (200 µM) - 5 µL, “Bottom-strand” DNA oligo (200 µM)- 5 µL, 10X Oligo Annealing Buffer - 2 µL, DNase/RNase-Free Water - 8 µL which should make a total volume of 20 µL.
2. If reannealing the lacZ ds control oligo, centrifuge its tube briefly (approximately 5 seconds), then transfer the contents to a separate 0.5 mL sterile microcentrifuge tube.
3. Incubate the reaction at 95 degrees C for 4 minutes.
4. Remove the tube containing the annealing reaction from the water bath or the heat block, and set it on your laboratory bench.
5. Allow the reaction mixture to cool to room temperature for 5-10 minutes. The single-stranded oligos will anneal during this time.
6. Place the sample in a microcentrifuge and centrifuge briefly (approximately 5 seconds). Mix gently.
7. Remove 1 µL of the annealing mixture and dilute the ds oligo as directed.
8. Store the remainder of the 50 µM ds oligo mixture at -20 degrees C.
You can verify the integrity of your annealed ds oligo by agarose gel electrophoresis, if desired.

What do I need to order to use your pENTR/U6 entry vector or pENTR/H1/TO vector?

You will need a double-stranded oligo that encodes the shRNA of interest to be cloned into one of the above-mentioned vectors. Use our RNAi Designer to design and synthesize two complementary single-stranded DNA oligonucleotides, with one encoding the shRNA of interest.

What does TO stand for in the pENTR/H1/TO vector?

TO stands for tetracycline operator, as this entry vector contains elements required for tetracycline-inducible expression of the shRNA in mammalian cells. The presence of the Tet operator sequences enables the shRNA of interest to be expressed in a tetracycline-dependent manner, thereby making this an inducible system.

What is the difference between the H1 and the U6 promoters?

The BLOCK-iT Inducible H1 and U6 Entry Vector Kits use either the Pol III-dependent H1 or the U6 promoter, respectively. The H1 promoter is modified to contain two flanking tetracycline operator (TetO2) sites within the H1 promoter. This allows the shRNA expressed from this promoter to be regulated in cells that express the tetracycline repressor (TR) protein. Both the H1 and the U6 are Pol III type promoters; however, there may be some minor differences in their effectiveness, depending on the cell line used.

What vectors do you offer for shRNA?

We offer our pENTR/U6 (Cat. No. K494500) and pENTR/H1/TO (Cat. No. K492000) vectors for shRNA delivery. Both vectors are Gateway compatible and drive expression through either the U6 or H1/TO promoter, respectively. The pENTR/H1/TO vector is for inducible shRNA expression, while the pENTR/U6 can be used for constitutive expression. If you want to design shRNA oligos compatible with both vectors, select the pENTER/U6 vector.

What are the general features of shRNA?

Exogenous short hairpin RNAs can be transcribed by RNA Polymerase III (Paule and White, 2000) and generally contain the following structural features: A short nucleotide sequence ranging from 19-29 nucleotides derived from the target gene, followed by a short spacer of 4-15 nucleotides (i.e., loop) and a 19-29 nucleotide sequence that is the reverse complement of the initial target sequence. The resulting RNA molecule forms an intramolecular stem-loop structure that is then processed to an siRNA duplex by the Dicer enzyme.

What does shRNA stand for, and how does it work?

Short hairpin RNA (shRNA) is an artificially designed class of RNA molecules that can trigger gene silencing through interaction with cellular components common to the RNAi and miRNA pathways. Although shRNA is a structurally simplified form of miRNA, these RNA molecules behave similarly to siRNA in that they trigger the RNAi response by inducing cleavage and degradation of target transcripts (Brummelkamp et al., 2002; Paddison et al., 2002; Paul et al., 2002; Sui et al., 2002; Yu et al., 2002). An RNA Polymerase III (Pol III), such as U6 and H1, drives transcription of shRNA transcripts. shRNA hairpins are exported from the nucleus and processed by Dicer into the cytosol, resulting in siRNA.

Why is a Pol III type promoter used for BLOCK-iT shRNA?

For efficient shRNA expression, a Pol III type promoter is used. These Pol III promoters contain all of their essential elements upstream of the expressed RNA and terminate with a short polythymidine tract. Once the shRNA is expressed, it is transported from the nucleus and processed into siRNA in the cytoplasm by the enzyme Dicer. Dicer preferentially recognizes shRNAs generated from a Pol III promoter because they carry no 5' or 3' flanking sequences. The siRNAs enter into RISC complexes and generate an RNAi response in mammalian cells.

I performed stable selection but my antibiotic-resistant clones do not express my gene of interest. What could have gone wrong?

Here are possible causes and solutions:

Detection method may not be appropriate or sensitive enough:
- We recommend optimizing the detection protocol or finding more sensitive methods. If the protein is being detected by Coomassie/silver staining, we recommend doing a western blot for increased sensitivity. The presence of endogenous proteins in the lysate may obscure the protein of interest in a Coomassie/silver stain. If available, we recommend using a positive control for the western blot.
- Insufficient number of clones screened: Screen at least 20 clones.
- Inappropriate antibiotic concentration used for stable selection: Make sure the antibiotic kill curve was performed correctly. Since the potency of a given antibiotic depends upon cell type, serum, medium, and culture technique, the dose must be determined each time a stable selection is performed. Even the stable cell lines we offer may be more or less sensitive to the dose we recommend if the medium or serum is significantly different.
- Expression of gene product (even low level) may not be compatible with growth of the cell line: Use an inducible expression system.
- Negative clones may result from preferential linearization at a vector site critical for expression of the gene of interest: Linearize the vector at a site that is not critical for expression, such as within the bacterial resistance marker.

I used a mammalian expression vector but do not get any expression of my protein. Can you help me troubleshoot?

Here are possible causes and solutions:

- Try the control expression that is included in the kit
Possible detection problem:

- Detection of expressed protein may not be possible in a transient transfection, since the transfection efficiency may be too low for detection by methods that assess the entire transfected population. We recommend optimizing the transfection efficiency, doing stable selection, or using methods that permit examination of individual cells. You can also increase the level of expression by changing the promoter or cell type.
- Expression within the cell may be too low for the chosen detection method. We recommend optimizing the detection protocol or finding more sensitive methods. If the protein is being detected by Coomassie/silver staining, we recommend doing a western blot for increased sensitivity. The presence of endogenous proteins in the lysate may obscure the protein of interest in a Coomassie/silver stain. If available, we recommend using a positive control for the western blot. Protein might be degraded or truncated: Check on a Northern. Possible time-course issue: Since the expression of a protein over time will depend upon the nature of the protein, we always recommend doing a time course for expression. A pilot time-course assay will help to determine the optimal window for expression. Possible cloning issues: Verify clones by restriction digestion and/or sequencing.

I am using a mammalian expression vector that has the neomycin resistance gene. Can I use neomycin for stable selection in mammalian cells?

No; neomycin is toxic to mammalian cells. We recommend using Geneticin (a.k.a. G418 Sulfate), as it is a less toxic and very effective alternative for selection in mammalian cells.

Is it okay if my construct has an ATG that is upstream of the ATG in my gene of interest? Will it interfere with translation of my gene?

Translation initiation will occur at the first ATG encountered by the ribosome, although in the absence of a Kozak sequence, initiation will be relatively weak. Any insert downstream would express a fusion protein if it is in frame with this initial ATG, but levels of expressed protein are predicted to be low if there is a non-Kozak consensus sequence. If the vector contains a non-Kozak consensus ATG, we recommend that you clone your gene upstream of that ATG and include a Kozak sequence for optimal expression.

I am planning to generate a T-REx cell line using pcDNA6/TR. Can I perform a western blot using antibodies to TetR to assess whether the cell line is expressing enough of TetR? Do you offer an antibody to TetR?

We do not offer an anti-TetR antibody. Even though a western using an anti-TetR antibody can be used to screen out clones that do not express any TetR protein, it would not be the optimal way to screen for functional clones. Functional testing by performing a transient transfection with the lacZ expression control plasmid is recommended for this purpose, followed by picking a clone that shows lowest basal levels of expression of beta-galactosidase in the absence of tetracycline, and highest levels of beta-galactosidase expression upon addition of tetracycline.

Do you offer a GFP-expressing mammalian expression vector that I can use as a control to monitor my transfection and expression?

We offer pJTI R4 Exp CMV EmGFP pA Vector, Cat. No. A14146, which you can use to monitor your transfection and expression.

I am working with a mouse cell line and would like to express my gene at high levels using one of your vectors with the CMV promoter. Do you foresee any problems with this approach?

The CMV promoter is known to be downregulated over time in mouse cell lines. Hence, we recommend using one of our non-CMV vectors, such as those with the EF1alpha or UbC promoter, for long-term expression in mouse cell lines.

Do I need to include a consensus Kozak sequence when I clone my gene of interest into one of your mammalian expression vectors?

The consensus Kozak sequence is A/G NNATGG, where the ATG indicates the initiation codon. Point mutations in the nucleotides surrounding the ATG have been shown to modulate translation efficiency. Although we make a general recommendation to include a Kozak consensus sequence, the necessity depends on the gene of interest and often, the ATG alone may be sufficient for efficient translation initiation. The best advice is to keep the native start site found in the cDNA unless one knows that it is not functionally ideal. If concerned about expression, it is advisable to test two constructs, one with the native start site and the other with a consensus Kozak. In general, all expression vectors that have an N-terminal fusion will already have an initiation site for translation.

Do I need to include a ribosomal binding site (RBS/Shine Dalgarno sequence) or Kozak sequence when I clone my gene of interest?

ATG is often sufficient for efficient translation initiation although it depends upon the gene of interest. The best advice is to keep the native start site found in the cDNA unless one knows that it is not functionally ideal. If concerned about expression, it is advisable to test two constructs, one with the native start site and the other with a Shine Dalgarno sequence/RBS or consensus Kozak sequence (ACCAUGG), as the case may be. In general, all expression vectors that have an N-terminal fusion will already have a RBS or initiation site for translation.

Can the Clontech Tet-On system or Tet-Off system components be used with your T-REx tetracycline-regulated mammalian expression system?

No. The two systems are not compatible since they utilize different strategies for promoter regulation. The T-REx system is designed such that native E. coli tet-repressor protein molecules bind to specific tet-operator sequences (2X TO) just downstream of the TATA box in the full length CMV promoter in the expression vector. This binding keeps the promoter silent simply by preventing the normal transcription machinery from productive assembly at the TATA box. Incidentally, it is this full length CMV promoter region that permits higher induced expression levels relative to other systems.

The recombinant 'repressor' proteins utilized in Clontech's system are actually recombinant fusion proteins which also contain a potent transcriptional transactivator. The Clontech system places operator sequences 5' to the TATA box and relies upon the VP16 transactivator to promote transcription. These repressor-transactivator fusion constructs would have unpredictable and unreliable effects at the CMV promoter in our expression constructs. Additionally, the tet-repressor protein produced from the pCDNA6/TR construct in the T-REx system has no transactivation domain and so would exert little regulatory effect at the minimal promoter region (non-full length CMV) found in the Clontech response plasmids.

Can you tell me the difference between a Shine-Dalgarno sequence and a Kozak sequence?

Prokaryotic mRNAs contain a Shine-Dalgarno sequence, also known as a ribosome binding site (RBS), which is composed of the polypurine sequence AGGAGG located just 5’ of the AUG initiation codon. This sequence allows the message to bind efficiently to the ribosome due to its complementarity with the 3’-end of the 16S rRNA. Similarly, eukaryotic (and specifically mammalian) mRNA also contains sequence information important for efficient translation. However, this sequence, termed a Kozak sequence, is not a true ribosome binding site, but rather a translation initiation enhancer. The Kozak consensus sequence is ACCAUGG, where AUG is the initiation codon. A purine (A/G) in position -3 has a dominant effect; with a pyrimidine (C/T) in position -3, translation becomes more sensitive to changes in positions -1, -2, and +4. Expression levels can be reduced up to 95% when the -3 position is changed from a purine to pyrimidine. The +4 position has less influence on expression levels where approximately 50% reduction is seen. See the following references:

- Kozak, M. (1986) Point mutations define a sequence flanking the AUG initiator codon that modulates translation by eukaryotic ribosomes. Cell 44, 283-292.
- Kozak, M. (1987) At least six nucleotides preceding the AUG initiator codon enhance translation in mammalian cells. J. Mol. Biol. 196, 947-950.
- Kozak, M. (1987) An analysis of 5´-noncoding sequences from 699 vertebrate messenger RNAs. Nucleic Acids Res. 15, 8125-8148.
- Kozak, M. (1989) The scanning model for translation: An update. J. Cell Biol. 108, 229-241.
- Kozak, M. (1990) Evaluation of the fidelity of initiation of translation in reticulocyte lysates from commercial sources. Nucleic Acids Res. 18, 2828.

Note: The optimal Kozak sequence for Drosophila differs slightly, and yeast do not follow this rule at all. See the following references:

- Romanos, M.A., Scorer, C.A., Clare, J.J. (1992) Foreign gene expression in yeast: a review. Yeast 8, 423-488.
- Cavaneer, D.R. (1987) Comparison of the consensus sequence flanking translational start sites in Drosophila and vertebrates. Nucleic Acids Res. 15, 1353-1361.

I sequenced one of your vectors after PCR amplification and observed a difference from what is provided online (or in the manual). Should I be concerned?

Our vectors have not been completely sequenced. Your sequence data may differ when compared to what is provided. Known mutations that do not affect the function of the vector are annotated in public databases.

Are your vectors routinely sequenced?

No, our vectors are not routinely sequenced. Quality control and release criteria utilize other methods.

How was the reference sequence for your vectors created?

Sequences provided for our vectors have been compiled from information in sequence databases, published sequences, and other sources.

What is the consensus Kozak sequence and what is the function of the Kozak sequence?

Eukaryotic (and specifically mammalian) mRNA contains sequence information that is important for efficient translation. However, this sequence, termed a Kozak sequence, is not a true ribosome binding site, but rather a translation initiation enhancer. The Kozak consensus sequence is ACCAUGG, where AUG is the initiation codon. A purine (A/G) in position -3 has a dominant effect; with a pyrimidine (C/T) in position -3, translation becomes more sensitive to changes in positions -1, -2, and +4. Expression levels can be reduced up to 95% when the -3 position is changed from a purine to pyrimidine. The +4 position has less influence on expression levels where approximately 50% reduction is seen. See the following references:

Kozak, M. (1986) Point mutations define a sequence flanking the AUG initiator codon that modulates translation by eukaryotic ribosomes. Cell 44, 283-292.
Kozak, M. (1987) At least six nucleotides preceding the AUG initiator codon enhance translation in mammalian cells. J. Mol. Biol. 196, 947-950.
Kozak, M. (1987) An analysis of 5´-noncoding sequences from 699 vertebrate messenger RNAs. Nucleic Acids Res. 15, 8125-8148.
Kozak, M. (1989) The scanning model for translation: An update. J. Cell Biol. 108, 229-241.
Kozak, M. (1990) Evaluation of the fidelity of initiation of translation in reticulocyte lysates from commercial sources. Nucleic Acids Res. 18, 2828.

Note: The optimal Kozak sequence for Drosophila differs slightly, and yeast do not follow this rule at all. See the following references:

Romanos, M.A., Scorer, C.A., Clare, J.J. (1992) Foreign gene expression in yeast: a review. Yeast 8, 423-488.
Cavaneer, D.R. (1987) Comparison of the consensus sequence flanking translational start sites in Drosophila and vertebrates. Nucleic Acids Res. 15, 1353-1361.


For Research Use Only. Not for use in diagnostic procedures.