Learn More
Invitrogen™ One Shot™ BL21(DE3)pLysS Chemically Competent E. coli
Description
One Shot BL21(DE3)pLysS Chemically Competent E. coli cells are ideal for use with bacteriophage T7 promoter-based expression systems such as pRSET and pET. The pLysS plasmid carried by the BL21(DE3) pLysS strain produces T7 lysozyme, an inhibitor of T7 RNA polymerase, to reduce basal level expression of the gene of interest. pLysS confers resistance to chloramphenicol (CamR) and contains the p15A origin. This origin allows pLysS to be compatible with plasmids containing the ColE1 or pMB1 origin (i.e. pUC- or pBR322- derived plasmids). Recombinant proteins that are nontoxic to E. coli are generally expressed at higher levels in BL21(DE3) cells compared to the BL21(DE3) pLysS strain with tighter expression control. BL21(DE3)pLysS is recommended for use when expressing toxic genes.
The BL21 (DE3) E. coli strain is one of the most popular host strains to produce recombinant proteins. This strain had been engineered to carry the λDE3 lysogen that contains T7 RNA polymerase gene under control of the lacUV5 promoter. IPTG is required to induce expression of the T7 RNA polymerase cascade system that in turn transcribe T7 promoter regulated target gene.
BL21(DE3) is a derivative from the E. coli B strain that does not contain the ion protease and is also deficient in the outer membrane protease OmpT. The lack of two key proteases reduces degradation of heterologous proteins expressed in the strains. Additionally, several other strain features make them especially suited for protein production, namely fast growth in minimal medium, a lower acetate production when grown in high glucose media, and an ability to reach high cell density. One Shot BL21(DE3)pLysS Chemically Competent E. coli cells can achieve transformation efficiency of >1 x 108 cfu/μg of control DNA.
One Shot BL21(DE3)pLysS Chemically Competent E. coli cells offer:
• Transformation efficiency of >1 x 108 cfu/μg control plasmid DNA
• Ion and OmpT protease deficiency that reduces degradation of recombinant protein
• IPTG-inducible lacUV5 promoter that controls expression of the T7 RNA polymerase
• pLysS (CamR) plasmid for reduced basal expression in uninduced cells
• hsdS mutation that allows efficient transformation of unmethylated DNA
• Fast growth in minimal medium and ability to reach high cell density
Easy-to-use One Shot format
BL21(DE3)pLysS Chemically Competent E. coli cells are supplied in the convenient, single-reaction One Shot format. The single-tube, single-use format allows all steps of the transformation protocol, up to plating, to take place in the same tube, thereby helping save time and to prevent contamination.
Genotype
F–ompT hsdSB (rB–, mB–) gal dcm (DE3) pLysS(CamR)
Find the strain and format that you need
• We offer other E. coli strains for protein expression.
• For expression of toxic proteins, consider BL21 AI One Shot Chemically Competent E. coli (Cat. No. C607003).
• BL21 Star (DE3) and other strains are available in MultiShot formats for high throughput applications.
• We offer a variety of systems for the expression of recombinant proteins in E. coli. The Champion pET expression system provides the highest level of protein production available in any expression system.
Specifications
Specifications
| Product Type | Chemically Competent Cells |
| Contains F' Episome | No |
| Improves Plasmid Quality | No |
| Cloning Methylated DNA | No |
| Transformation Efficiency Level | Medium Efficiency (1 x 108 to 1 x 109 cfu/μg) |
| Content And Storage | • Chemically Competent cells (21 x 50 μL); store at –80°C • pUC19 Control DNA (1 x 50 μL); store at –80°C • S.O.C. Medium (6 mL); store at 4°C or room temperature |
| Antibiotic Resistance Bacterial | Yes (Chloramphenicol) |
| Blue/White Screening | No |
| High-throughput Compatibility | Low |
| Preparing Unmethylated DNA | Yes (dcm) |
| Show More |
Frequently Asked Questions (FAQs)
Several precautions may be taken to prevent problems resulting from basal level expression of a toxic gene of interest. These methods all assume that the T7-based or Champion-based expression plasmid has been correctly designed and created.
- Propagate and maintain your expression plasmid in a strain that does not contain T7 RNA polymerase (i.e., DH5α).
- If using BL21 (DE3) cells, try growing cells at room temperature rather than 37 degrees C for 24-48 hr.
- Perform a fresh transformation using a tightly regulated E. coli strain, such as BL21-AI cells.
- After following the transformation protocol, plate the transformation reaction on LB plates containing 100 µg/mL ampicillin and 0.1% glucose. The presence of glucose represses basal expression of T7 RNA polymerase.
- Following transformation of BL21-AI cells, pick 3 or 4 transformants and inoculate directly into fresh LB medium containing 100 µg/mL ampicillin or 50 µg/mL carbenicillin (and 0.1% glucose, if desired). When the culture reaches an OD600 of 0.4, induce expression of the recombinant protein by adding L-arabinose to a final concentration of 0.2%.
- When performing expression experiments, supplement the growth medium with 0.1% glucose in addition to 0.2% arabinose.
- Try a regulated bacterial expression system such as our pBAD system.
Typically, if you see 1-2 dominant bands, translation stopped prematurely due to codon usage bias. With degradation, you usually see a ladder of bands. With degradation, you can try using a protease inhibitor and add it to the lysis buffer to help prevent degradation. If degradation is the issue, a time point experiment can be done to determine the best time to harvest the cells.
If you are having a solubility issue, try to decrease the temperature or decrease the amount of IPTG used for induction. You can also try a different, more stringent cell strain for expression. Adding 1% glucose to the bacterial culture medium during expression can also help.
- Inoculate from fresh bacterial cultures, since higher protein yields are generally obtained from a fresh bacterial colony.
- Check the codon usage in the recombinant protein sequence for infrequently used codons. Replacing the rare codons with more commonly used codons can significantly increase expression levels. For example, the arginine codons AGG and AGA are used infrequently by E. coli, so the level of tRNAs for these codons is low.
- Add protease inhibitors, such as PMSF, to buffers during protein purification. Use freshly made PMSF, since PMSF loses effectiveness within 30 min of dilution into an aqueous solution.
- If you are using ampicillin for selection in your expression experiments, you may be experiencing plasmid instability due to the absence of selective conditions. This occurs as the ampicillin is destroyed by β-lactamase or hydrolyzed under the acidic media conditions generated by bacterial metabolism. You may want to substitute carbenicillin for ampicillin in your transformation and expression experiments.
- The recombinant protein may be toxic to bacterial cells. Try a tighter regulation system for competent cell expression such as BL21-AI. You may also consider trying a different expression system such as the pBAD system.
This typically occurs when your gene of interest is toxic. Try using a tighter regulation system, such as BL21 (DE3) (pLysS) or BL21 (DE3) (pLysE), or BL21(AI).
For Research Use Only. Not for use in diagnostic procedures.