Learn More
Invitrogen™ Mifepristone
Catalog No. H11001
Encompass
Change view
Click to view available options
Quantity:
100 μg
Catalog No. Quantity
H11001 100 μg
1 options
Catalog No. H11001 Supplier Invitrogen™ Supplier No. H11001
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Includes
100μg mifepristone is provided lyophilized.
Progesterone antagonist that activates transcription
  • Subphysiological levels of steroid compound are used to induce expression from very low basal to high induced levels
  • Formulated for use in GeneSwitch™ System
  • Lypholized
Mammalian Expression, Protein Expression, Proteins, Expression, Isolation and Analysis, Regulated Expression
Specifications
Format Lyophilized
Product Type Mifepristone
Content And Storage 100 μg mifepristone is provided lyophilized. Store at -20°C. Mifepristone is guaranteed stable for 6 months when properly stored.
Quantity 100 μg
For Research Use Only. Not for use in diagnostic procedures.