missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MAX Efficiency™ DH5α Competent Cells

Catalog No. 18258012
Encompass
Change view
Click to view available options
Quantity:
5 x 200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18258012 5 x 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18258012 Supplier Invitrogen™ Supplier No. 18258012
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Max Efficiency DH5α Competent Cells: 5 vials, 200μL each (total of 1mL), pUC19 DNA (0.01μg/mL): 1 vial, 100μL, S.O.C. medium: 2 bottles, 6mL each

MAX Efficiency™ DH5α Competent Cells are a well-known, versatile strain that is used in many everyday cloning applications. Prepared by a proprietary modification of the Hanahan method, these cells are suitable for high efficiency transformations in applications where low amount of DNA is used or to construct libraries using plasmid-derived vectors.

MAX Efficiency™ DH5α Competent Cells are a well-known, versatile strain that is used in many everyday cloning applications. Prepared by a proprietary modification of the Hanahan method, these cells are suitable for high efficiency transformations in applications where low amount of DNA is used or to construct libraries using plasmid-derived vectors. In addition to supporting blue/white screening, recA1 and endA1 mutations in DH5α cells increase insert stability and improve the quality of plasmid DNA prepared from minipreps.

Benefits of MAX Efficiency DH5α Competent Cells:
• Versatile strain for everyday cloning applications
• High quality plasmid preparations
• Useful for generating plasmid libraries 

DH5α competent cells for high transformation efficiency
The Φ80lacZΔM15 marker provides α-complementation of the β-galactosidase gene from pUC or similar vectors, and can be used for blue/white screening of colonies on bacterial plates containing Bluo-gal or X-gal. DH5α cells are capable of being transformed efficiently with large plasmids, and can also serve as a host for the M13mp cloning vectors if a lawn of DH5αF′IQ (Cat. No. 18288019) (or similar) is provided to allow plaque formation.

Features of MAX Efficiency DH5α Competent Cells include:
• Transformation efficiency of >1 x 109 transformants/μg plasmid DNA
• High plasmid yield from the DH5α (endA1) E. coli strain 
• Blue/white screening capable (lacZΔM15)
• Greater insert stability (recA1)

Genotype:
F- Φ80lacZΔM15 Δ(lacZYA-argF) U169 recA1 endA1 hsdR17 (rk-, mk+) phoA supE44 λ-thi-1 gyrA96 relA1 

Find the strain and format that fits you needs
• DH5α strain and its derivatives available in with a variety of transformation efficiencies and in both electrocompetent and chemically competent formats. 
• ElectroMAX™ DH5α-E Competent Cells (Cat. No. 11319019) are engineered for electroporation to achieve high transformation efficiency of >1 x 1010.
• DH5α T1R competent cells are available in MultiShot format for high throughput applications.

Order Info

Shipping Condition: Dry Ice

Specifications

Product Type Chemically Competent Cells
Contains F' Episome No
Improves Plasmid Quality Yes (endA1)
Cloning Methylated DNA No
Transformation Efficiency Level High Efficiency (>1 x 109 cfu/μg)
Content And Storage • Max Efficiency DH5α Competent Cells (5 x 200 μL)
Store Competent Cells at –80°C.

• pUC19 DNA (100 μL at 0.01 μg/mL)
Store pUC19 DNA at –20°C.

• S.O.C. Medium (2 x 6 mL)
Store S.O.C. Medium at 4°C or room temperature.

Antibiotic Resistance Bacterial No
Cloning Unstable DNA Not suitable for cloning unstable DNA
Blue/White Screening Yes (lacZΔM15)
High-throughput Compatibility Low
Preparing Unmethylated DNA No
Reduces Recombination Yes (recA1)
Shipping Condition Dry Ice
T1 Phage - Resistant (tonA) No
Species E. coli (K12)
Format Tube
Product Line MAX Efficiency
Quantity 5 x 200 μL
Show More Show Less
I am trying to clone an insert that is supposedly pretty toxic. I used DH5? and TOP10 cells for the transformation and got no colonies on the plate. Do you have any suggestions for me?

If the insert is potentially toxic to the host cells, here are some suggestions that you can try:

- After transforming TOP10 or DH5? cells, incubate at 25-30°C instead of 37°C. This will slow down the growth and will increase the chances of cloning a potentially toxic insert.
- Try using TOP10F' cells for the transformation, but do not add IPTG to the plates. These cells carry the lacIq repressor that represses expression from the lac promoter and so allows cloning of toxic genes. Keep in mind that in the absence of IPTG, blue-white screening cannot be performed.
- Try using Stbl2 cells for the transformation.

How do you recommend that I prepare my DNA for successful electroporation of E. coli?

For best results, DNA used in electroporation must have a very low ionic strength and a high resistance. A high-salt DNA sample may be purified by either ethanol precipitation or dialysis.

The following suggested protocols are for ligation reactions of 20ul. The volumes may be adjusted to suit the amount being prepared.

Purifying DNA by Precipitation: Add 5 to 10 ug of tRNA to a 20ul ligation reaction. Adjust the solution to 2.5 M in ammonium acetate using a 7.5 M ammonium acetate stock solution. Mix well. Add two volumes of 100 % ethanol. Centrifuge at 12,000 x g for 15 min at 4C. Remove the supernatant with a micropipet. Wash the pellet with 60ul of 70% ethanol. Centrifuge at 12,000 x g for 15 min at room temperature. Remove the supernatant with a micropipet. Air dry the pellet. Resuspend the DNA in 0.5X TE buffer [5 mM Tris-HCl, 0.5 mM EDTA (pH 7.5)] to a concentration of 10 ng/ul of DNA. Use 1 ul per transformation of 20 ul of cell suspension.

Purifying DNA by Microdialysis: Float a Millipore filter, type VS 0.025 um, on a pool of 0.5X TE buffer (or 10% glycerol) in a small plastic container. Place 20ul of the DNA solution as a drop on top of the filter. Incubate at room temperature for several hours. Withdraw the DNA drop from the filter and place it in a polypropylene microcentrifuge tube. Use 1ul of this DNA for each electrotransformation reaction.

You offer competent cells in Subcloning Efficiency, Library Efficiency and MAX Efficiency. How do these differ?

There are a few exceptions, but in general the difference is in guaranteed transformation efficiency as follows:

Subcloning Efficiency cells are guaranteed to produce at least 1.0 x 10E6 transformants per µg of transformed pUC19 or pUC18 supercoiled plasmid
Library Efficiency cells are guaranteed to produce at least 1.0 x 10E8 transformants per µg pUC19 or pUC18 DNA
MAX Efficiency cells are guaranteed to produce at least 1.0 x 10E9 transformants per µg pUC19 or pUC18 DNA

What are the advantages of using TOP10 over DH5alpha cells for cloning?

The main advantage is that TOP10 cells have mutations in the mcrA, mcrB and mrr genes which encode restriction systems for methylated DNA. This means that you can clone highly methylated DNA derived from such sources as mammalian and plant cells, and it will not be degraded after transformation.

Do any Invitrogen competent cells contain DMSO in the freezing medium?

Yes, several of our competent cells products are frozen with DMSO. The presence of DMSO (dimethylsulfoxide) will generally be indicated in the MSDS files if you have a question about a particular product, but here is a list of commonly used products that are known to have DMSO in the freezing buffer:

One Shot OmniMAX 2 T1 Phage Resistant Cells, Cat. No. C8540-03

One Shot INV?F' Chemically Competent Cells, Cat. No. C2020-03 and C2020-06

One Shot MAX Efficiency DH5?-T1 Chemically Competent Cells, Cat. No. 12297-016

MAX Efficiency DH5?-T1 Phage Resistant Cells, Cat. No. 12034-013

MAX Efficiency DH5? Chemically Competent Cells, Cat. No. 18258-012

Library Efficiency DH5? Chemically Competent Cells, Cat. No. 18263-012

MAX Efficiency DH5? F'IQ Cells, Cat. No. 18288-019

MAX Efficiency Stbl2Chemically Competent Cells, Cat. No. 10268-019

Is S.O.C. medium absolutely required when recovering competent bacterial cells during transformation?

Many media can be used to grow transformed cells, including standard LB, SOB or TB broths. However, S.O.C. is the optimal choice for recovery of the cells before plating. The nutrient-rich formula with added glucose is often important for obtaining maximum transformation efficiencies.

Why is it necessary to dilute ligated DNA products before adding them to competent bacterial cells?

Components of the ligation reaction (enzymes, salts) can interfere with transformation, and may reduce the number of recombinant colonies or plaques. We recommend a five-fold dilution of the ligation mix, and adding not more than 1/10 of the diluted volume to the cells. For best results, the volume added should also not exceed 10% of the volume of the competent cells that you are using.

When should DMSO, formamide, glycerol and other cosolvents be used in PCR?

Cosolvents may be used when there is a failure of amplification, either because the template contains stable hairpin-loops or the region of amplification is GC-rich. Keep in mind that all of these cosolvents have the effect of lowering enzyme activity, which will decrease amplification yield. For more information see P Landre et al (1995). The use of co-solvents to enhance amplification by the polymerase chain reaction. In: PCR Strategies, edited by MA Innis, DH Gelfand, JJ Sninsky. Academic Press, San Diego, CA, pp. 3-16.

Additionally, when amplifying very long PCR fragments (greater than 5 kb) the use of cosolvents is often recommended to help compensate for the increased melting temperature of these fragments.


For Research Use Only. Not for use in diagnostic procedures.