missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Lipofectamine™ Transfection Reagent

Catalog No. p-4924039
Encompass
Change view
Click to view available options
Quantity:
0.3 mL
1 mL
4 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18324010 0.3 mL
18324012 1 mL
18324020 4 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18324010 Supplier Invitrogen™ Supplier No. 18324010
Only null left

Includes

Contains one vial of 1mL.

Lipofectamine™ Transfection Reagent is one of our first-generation, cationic-lipid transfection reagents formulated for the transfection of DNA into eukaryotic cells.

Lipofectamine™ Transfection Reagent is one of our first-generation, cationic-lipid transfection reagents formulated for the transfection of DNA into eukaryotic cells. Lipofectamine™ Transfection Reagent is a trusted, value reagent with many years of cited use in thousands of publications and with many cell lines.

With Lipofectamine™ Transfection Reagent, you'll get:

  • A reagent proven to work in high-throughput applications
  • A reliable reagent for establishing stable cell lines
  • A reagent that works well with PLUS™ reagent for higher protein expression

A proven technology

Our Lipofectamine™-brand reagents have been recognized as the most-cited transfection reagent family, with tens of thousands of citations since launch in 1993. Lipofectamine™ Transfection Reagent is no exception as a trusted reagent for a wide range of cell lines. When used in combination with PLUS™ Reagent, Lipofectamine™ Transfection Reagent has been shown to work well with cells such as BHK-21, NIH 3T3, COS-1, fibroblasts, keratinocytes, HT-29, MRC-5, and SK-BR3.

A 'value' alternative for plasmid DNA

With such a legacy comes a commitment to providing the best reagents for all scientists and labs, regardless of their funding capabilities. Lipofectamine™ Transfection Reagent is a value alternative for transfection of plasmid DNA into eukaryotic cells with a simplified protocol. For optimal results in a wider range of plasmid DNA transfection conditions, we recommend Lipofectamine™ LTX Reagent, due to its optimal balance of potency & low-cytotoxicity. Lipofectamine™ 2000 Transfection Reagent is recommended if you are transfecting a broad range of cell lines or want to transfect RNA.

Order Info

Shipping Condition: Wet Ice

Specifications

Content And Storage Each unit contains one or multiple vials of reagent. Store at 2°C to 8°C.
Cell Type Primary Cells, Hard-to-Transfect Cells
Format 6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks
Sample Type Plasmid DNA
Transfection Technique Lipid-based Transfection
For Use With (Application) Transfection
High-throughput Compatibility High-throughput Compatible
Product Line Lipofectamine
Product Type Transfection Reagent
Quantity 0.3 mL
Serum Compatible No
Shipping Condition Wet Ice
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.