missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Lipofectamine™ RNAiMAX Transfection Reagent
GREENER_CHOICE

Catalog No. p-4924036
Encompass
Change view
Click to view available options
Quantity:
0.1 mL
0.3 mL
0.75 mL
1.5 mL
15 mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13778075 0.75 mL
13778150 1.5 mL
13778100 0.1 mL
13778030 0.3 mL
13778500 15 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 13778075 Supplier Invitrogen™ Supplier No. 13778075
Only null left

Achieve effective RNAi transfection with Lipofectamine RNAiMAX Transfection Reagent across a wide range of cell types. This lipid-based transfection reagent is optimized for siRNA and miRNA transfection, offering high delivery efficiency and low cellular toxicity, enabling effective gene silencing.

Lipofectamine RNAiMAX reagent is a leading transfection reagent designed specifically for RNA interference applications. It efficiently delivers siRNA and miRNA into a wide variety of cell types, ensuring high transfection efficiency and effective gene knockdown. The proprietary cationic lipid formulation minimizes cytotoxicity, making it suitable for sensitive cells and long-term studies. Lipofectamine RNAiMAX reagent is optimized for consistent performance, providing researchers with reliable results in gene silencing experiments. Its ease of use and compatibility with high-throughput screening make it an essential tool for basic biology research and functional genomics.

Features of Lipofectamine RNAiMAX Transfection Reagent

  • Optimized for RNAi—Specifically formulated for RNA interference applications, maximizing siRNA and miRNA transfection and gene silencing.
  • Superior siRNA transfection efficiency—requires lower siRNA concentrations for effective gene knockdown
  • miRNA reagent compatible—provides superior transfection efficiencies for miRNA mimics and inhibitors
  • Minimal cytotoxicity—flexible 10-fold concentration range of the siRNA transfection reagent with low cellular impact
  • Broad cell type compatibility—versatile use in a wide range of cell lines, including hard-to-transfect cells
  • Simple and rapid protocol—easy-to-follow steps that save time and effort in the lab for consistent and reproducible results

High knockdown in a wide range of cells

Lipofectamine RNAiMAX Transfection Reagent is an effective siRNA transfection reagent suitable for a wide range of cell types, including primary cells (see figure). It achieves high-efficiency siRNA transfection, leading to significant gene knockdown necessary for convincing gene silencing results.

Simple, high-throughput-ready transfections

Mix Lipofectamine RNAiMAX Transfection Reagent with siRNA, add to your cells, incubate, and measure gene knockdown. Its simplicity and speed, combined with high transfection efficiency, make it ideal for high-throughput siRNA transfection workflows. Whether it's a forward or reverse transfection protocol, siRNA transfection conditions can be easily optimized for automated or robotic systems.

Order Info

Shipping Condition: Qualified for Ambient or Wet Ice

Specifications

Content And Storage Contains one vial (0.75 ml) Lipofectamine™ RNAiMAX Transfection Reagent. Store at 2–8°C. Do not freeze.
Cell Type Established Cell Lines, Stem Cells, Primary Cells, Hard-to-Transfect Cells
Format 6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks
Sample Type Synthetic siRNA, siRNA, miRNA
Transfection Technique Lipid-based Transfection
For Use With (Application) Transfection
High-throughput Compatibility High-throughput Compatible
Product Line Lipofectamine
Product Type Transfection Reagent
Quantity 0.75 mL
Serum Compatible Yes
Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.