missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ INVSc1 S. cerevisiae Yeast Strain

Catalog No. C81000
Encompass
Change view
Click to view available options
Quantity:
1 Stab
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C81000 1 Stab
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C81000 Supplier Invitrogen™ Supplier No. C81000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Fast-growing strain ideal for expression

The S. cerevisiae strain INVSc1 is a fast-growing strain ideal for expression. INVSc1 has the following genotype:

INVSc1: MATa his3D1 leu2 trp1-289 ura3-52 MAT his3D1 leu2 trp1-289 ura3-52

Specifications

Product Type Yeast Strain
Bacterial or Yeast Strain INVSc1
Cell Line Yeast
Species S. cerevisiae
Quantity 1 Stab
What is the trp1-289 mutation in the INVSc1 strain?

The trp1-289 mutation is a point mutation in the TRP-1 gene that causes this strain to be auxotrophic for tryptophan (i.e., tryptophan is required in growth media for this strain). There are many vectors that contain a wild-type copy of the TRP1 gene that will complement the trp1-289 mutant phenotype and can therefore be used as a selectable marker for such vectors. The phenotype of a trp1-289 mutant and a trp1 complete deletion mutant are similar, but strains with the trp1 complete deletion, unlike trp1-289, will not induce well with galactose. Therefore, when galactose induction is used, it is better to use a trp1-289 mutant. However, since trp1-289 mutants revert with a detectable frequency, it is important to verify your clones.

Will S. cerevisiae grow differently using galactose instead of glucose as a carbon source?

S. cerevisiae can grow using either or both mechanisms of carbon metabolism. The balance between the two is different for glucose vs. galactose as a carbon source. Under ideal conditions, S. cerevisiae grows slower on galactose than on glucose, because production of glucose-6-P from galactose is rate limiting. (gal -> gal-1-P -> glu-1-P -> glu-6-P). Under non-ideal conditions (low oxygen, as in the center of a colony or a culture without really good oxygen feed), it becomes even worse because cells grown on galactose are using more respiration than fermentation relative to cells grown on glucose. Low oxygen makes fermentation more necessary, which cells growing on galactose are not good at.

Should D-raffinose be used as carbon source for yeast prior to galactose induction? Does L-raffinose work?

As with other sugars (e.g. glucose), D-raffinose is the biologically active carbon source for yeast. Pure L-raffinose will not work.

What is an appropriate innoculum amount to begin a galactose induction experiment? How does raffinose affect the time course of galactose induction?

The suggested initial cell density for galactose induction is 1 to 5 X 10E6 cells/ml . The cells are allowed to divide one or two times and then induced with galactose. Galactose induction is best in log phase and the culture will probably approach static phase at 1 to 4 X 10E7 cells/ml. Induction of cells maintained in raffinose may begin in 15 to 30 minutes whereas induction of cells maintained in glucose may not first occur for an hour or more. Peak expression will often occur in 2 - 4 hours so time points should be taken every hour (or every other hour) for up to 10 hours. When using raffinose maintained cells, the induction is much faster than induction of glucose maintained cells. Maximal expression levels remain the same.

What are some of the common types of auxotrophic markers in yeast?

The following are commonly employed auxotrophic markers:

1) his3Δ1: Histidine requiring strain (from gene disruption) with a deletion in locus 1. The his3 denotes the disruption of the HIS3 gene. The Δ1 is a deletion that has been engineered to decrease the recombination between the incoming plasmid DNA and the chromosomal site.
2) leu2: Leucine requiring strain due to the disruption of the LEU2 gene.
3) trp1-289: Tryptophan requiring strain, developed from gene disruption and a further point mutation to decrease the recombination between the incoming plasmid DNA and the chromosomal site.
4) ura3-52: Uracil requiring.

For more detail on types and methods of gene disruption in yeast refer to METHODS IN ENZYMOLOGY Vol. 194.

I sequenced one of your vectors after PCR amplification and observed a difference from what is provided online (or in the manual). Should I be concerned?

Our vectors have not been completely sequenced. Your sequence data may differ when compared to what is provided. Known mutations that do not affect the function of the vector are annotated in public databases.

Are your vectors routinely sequenced?

No, our vectors are not routinely sequenced. Quality control and release criteria utilize other methods.

How was the reference sequence for your vectors created?

Sequences provided for our vectors have been compiled from information in sequence databases, published sequences, and other sources.


For Research Use Only. Not for use in diagnostic procedures.