missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GripTite™ 293 MSR Cell Line

Catalog No. R79507
Encompass
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R79507 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R79507 Supplier Invitrogen™ Supplier No. R79507

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

The GripTite 293 MSR Cell Line is provided as two tubes of a frozen stock of 3 x 106 cells, each in 1mL of 90% complete media and 10% DMSO. Liquid Geneticin Selective Antibiotic (20mL) is supplied as a 50mg/mL solution. Versene (100mL) contains 0.2g/L EDTA 4Na in phosphate-buffered saline. 1X concentration is 0.2g/L, 0.53mM.

A genetically engineered Human Embryonic Kidney (HEK 293) cell line that expresses the human macrophage scavenger receptor

  • Strongly adheres to standard tissue culture plates for dependable results
  • Show the same fast cell growth, high transfection efficiency, and high-level expression characteristics of the parental 293-H cells
  • Unlike most 293 cells, these cells don't wash away during the repeated manipulations of routine tissue culture or plate washing protocols
  • Superior adherence enables reliable reproduction of ligand-binding, enzymatic, or immunofluorescence assay results
  • The GripTite293 MSR cells enhance performance in high-throughput cell-based, as well as standard tissue culture, applications
  • No need for the poly-lysine coated plates typically used in high-throughput analysis: withstand automated plate washing and protocols using liquid handling robots on standard tissue culture plates without significant cell loss

Antibody and Cell Culture Production, Bioproduction, Cell Culture/Mammalian Cell Culture, Mammalian Expression, Mammalian Expression Cell Lines, Mammalian Protein Production, Plasmid Transfection, Protein Expression, Proteins, Expression, Isolation and Analysis, RNAi Transfection, Synthetic siRNA Transfection, Transfection, shRNA and miR RNAi Plasmid Transfection

Specifications

Content And Storage The GripTite™ 293 MSR Cell Line is provided as two tubes of a frozen stock of 1 x 107 cells, each in 1 ml of 90% complete media and 10% DMSO. Store in liquid nitrogen. Liquid Geneticin™ Selective Antibiotic (20 ml) is supplied as a 50 mg/ml solution. Store at -20°C or 2°C to 8°C. Versene (100 ml) contains 0.2 g/L EDTA 4Na in phosphate-buffered saline. 1X concentration is 0.2 g/L=0.53 mM. Store at 2°C to 8°C. Guaranteed stable for 6 months when properly stored.
Cell Line 293 GripTite™ MSR
Species Human
Concentration 1X
Quantity 1 kit
Product Line GripTite

For Research Use Only. Not for use in diagnostic procedures.