missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FreeStyle™ MAX 293 Expression System

Catalog No. K900010
Encompass
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K900010 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K900010 Supplier Invitrogen™ Supplier No. K900010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rapid and high-yield mammalian protein production

FreeStyle MAX Reagent is a novel cationic lipid-based reagent designed to transfect FreeStyle cells with high efficiency. The FreeStyle MAX Expression System combines GIBCO™ FreeStyle Media, FreeStyle MAX Reagent, and either FreeStyle CHO-S cells or FreeStyle 293 cells. The only equipment required to use the FreeStyle MAX System is a CO2 incubator and a platform shaker with shake flasks or a spinner culture

FreeStyle MAX System provides:

  • Easy, rapid protocol for transient transfection of CHO and 293 cells in suspension cultures
  • Functional proteins (up to milligram quantities) with all post-translational modifications produced in one week
  • Simplified downstream purification of secreted proteins with FreeStyle serum-free medium
  • Straightforward protocol for production scale-up
  • Replace time-consuming stable cell line generation with large-scale functional protein production in one week

Antibody and Cell Culture Production, Bioproduction, Cell Culture, Mammalian Expression, Mammalian Protein Production, Plasmid Transfection, Protein Expression, Proteins, Expression, Isolation and Analysis, Transfection, Transient Protein Production and High-Throughput Screening

Specifications

Cell Line FreeStyle™ 293-F Cells
Classification Animal Origin-Free, Serum-Free
Content And Storage Store FreeStyle cells in liquid nitrogen. Store FreeStyle MAX Reagent at 2–8°C. Store FreeStyle CHO Expression Medium, FreeStyle 293 Expression Medium, and OptiPRO SFM at 2–8°C in the dark. Store the pCMV SPORT at -20°C. Stability is guaranteed for 6 months under the recommended storage conditions. Please refer to the manual for additional storage information.
Delivery Type Transfection
Culture Type Suspension Cell Culture
Cell Type 293-derived Cells
Transfection Technique Lipid-based Transfection, Transient Transfection
Expression Mechanism Cell-Based Expression
Expression System Mammalian
For Use With (Application) Protein Expression
Key Functions Protein Production
Product Line Freestyle
Product Type Expression System
Quantity 1 Kit
Show More Show Less
What is the main difference between the FreeStyle Max 293 Expression System and the FreeStyle 293 Expression System?

The two systems have the same components except for the transfection reagent and the medium supplied for formation of transfection complexes. The FreeStyle Max 293 Expression System contains FreeStyle Max Reagent and OptiPRO SFM (serum-free medium that is devoid of animal-origin components), whereas the FreeStyle 293 Expression System contains 293fectin Transfection Reagent and Opti-MEM I Reduced Serum Medium (serum-free medium but not animal-origin free).

What is the recommended ratio of DNA and transfection reagent when transfecting Freestyle cells?

The optimal amount will vary, but we recommend starting with a 1:1 ratio of DNA and lipid. For example, 1.25 ug DNA and 1.25 ul of the reagent for one ml of suspension cells.

Which vectors do you recommend for use with FreeStyle system?

Generally, any mammalian expression vector should be compatible with the FreeStyle expression system. Some of the more popular choices include pcDNA3.3-TOPO TA vector (Cat. No. K8300-01), pcDNA3.4-TOPO TA vector (Cat. no. A14697), pcDNA3.2 DEST vector (Cat. No. 12489018), pCEP vector (Cat. No. V044-50), or pcDNA4/HisMax vector (Cat. No. K864-20).

How does FreeStyle MAX reagent compare to Lipofectamine 2000 and 293fectin reagents?

Lipofectamine 2000 is designed for adherent cells. FreeStyle and 293fectin reagents are designed for suspended Hek293 and CHO cells. For transfecting 293 suspension cells, FreeStyle MAX reagent yields transfection efficiency comparable to Lipofectamine 2000 and 293fectin reagents. However, Lipofectamine 2000 and 293fectin reagents do not work well for CHO suspension cells.

What's the transfection efficiency for FreeStyle MAX reagent?

In general, the transfection efficiency for FreeStyle MAX reagent is 65% when used with suspension CHO and 293 cells.

For Research Use Only. Not for use in diagnostic procedures.