missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Flp-In™ Core System

Catalog No. K601002
Encompass
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K601002 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K601002 Supplier Invitrogen™ Supplier No. K601002

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

The Flp-In Core Kit includes 20μg each of pFRT/lacZeo, pOG44, pcDNA5/FRT, and pcDNA5/FRT/CAT (expression control), and 2μg each of forward and reverse sequencing primers.

Contains necessary vectors that allow integration and expression of gene of interest in mammalian cells at specific genomic location

  • For introduction of Flp Recombination Target (FRT) site into genome of mammalian cell line of choice
  • Expression vector containing gene of interest is then integrated into genome via Flp recombinase-mediated DNA recombination at FRT site

Constitutive Expression, Mammalian Expression, Protein Expression, Proteins, Expression, Isolation and Analysis, Targeted Integration

Specifications

Constitutive or Inducible System Constitutive
Delivery Type Transfection
Promoter CMV
Product Type Flp-In Core System
Selection Agent (Eukaryotic) Zeocin™
Content And Storage The Flp-In™ Core Kit includes 20 μg each of pFRT⁄lacZeo, pOG44, pcDNA™5⁄FRT, and pcDNA™5⁄FRT⁄CAT (expression control), and 2 μg each of forward and reverse sequencing primers. Store at -20C.
Protein Tag Untagged
Cloning Method Restriction Enzyme/MCS
Quantity 1 kit
Vector pFRT
Product Line Flp-In, pcDNA
For Use With (Application) Targeted Integration
Show More Show Less
I performed the Flp-In reaction and obtained hygromycin-resistant clones which also turned out to be Zeocin antibiotic resistant and lacZ-positive. I am concerned that the Flp-In reaction has not worked. Can you explain what might be happening?

We have observed in-house that in cells where the FRT site has integrated into a very transcriptionally active locus in the host cell genome (seen more commonly in Flp-In CHO and Flp-In 293 cells but can also happen in Flp-In 3T3 cells and any other Flp-In host cell line), there is some "read-through" transcription and translation of the lacZ-Zeocin ORF subsequent to the Flp-In reaction, even though the lacZ-Zeocin ORF does not have a bonafide promoter and ATG. In such cases, the hygromycin-resistant clones would also be lacZ-positive and Zeocin antibiotic-resistant. To make sure that the integration is FRT site-specific and not random, we recommend doing a parallel control transfection with no pOG44 present. This should yield no surviving clones upon hygromycin selection, indicating that all the hygromycin-resistant clones obtained in the presence of pOG44 are indeed Flp recombinase-dependent and hence have the gene of interest integrated at the FRT site. Also, a Southern blot analysis of these clones will help verify that they do indeed have proper FRT integration of the gene of interest despite the expression of lacZ (although this is typically not necessary). After the Flp-In reaction, as long as you see hygromycin-resistant clones, we recommend that you select them and assay them for expression of your gene of interest.

I used the pcDNA5/FRT vector in your Flp-In 3T3 cell line and was initially able to express my gene of interest, but after a month or so, lost expression. Why is this?

The Flp-In 3T3 cell line is derived from NIH3T3 cells, which are mouse fibroblast cells. The CMV promoter is known to get silenced over time in murine cell lines and hence we would recommend using a Flp-In expression vector with a non-CMV promoter in these cells, such as the pEF5/FRT/V5-D-TOPO vector or the pEF5/FRT/V5-DEST vector.

I used the pFRT/lacZeo vector to generate my own Flp-In host cell line and then made my Flp-In expression cell line. I am seeing very poor expression of my gene of interest. Is there anything else I can try to improve expression?

Before giving up, we would suggest that you try using the pFRT/lacZeo2 vector to generate your host cell line. This vector contains a truncated version of the SV40 promoter driving the lacZ-Zeocin fusion. Use of this vector facilitates the isolation of clones that have integrated the vector near enhancer elements in the genome, thus resulting in higher levels of expression of the gene of interest.

What is the difference between the Jump-In and Flp-In systems?

The Jump-In system is PhiC31-integrase mediated and is a stable, targeted, and irreversible mammalian expression system. It consists of the Jump-In Fast system that involves a single integration step and the Jump-InTI (targeted integration) system that needs two integration steps, both of which are targeted and irreversible. In contrast, the Flp-In system is a stable, targeted mammalian expression system that is reversible. The first integration is random (integration of pFRT/lacZeo), and the second integration (integration of the Flp-In expression vector) is targeted but reversible.

Is multiple integration of the Flp-In expression construct possible? How do you screen for multiple integrants, and how stable is the Flp-In expression cell line?

In theory, one can get multiple integrations of the Flp-In expression construct—an FRT-specific integration event and a random, second-site integration. However, random integration is a relatively uncommon event. Limiting the amount of DNA in the transfection will reduce the chance of second-site integration. We have transfected 293 cells (lacking the FRT site) with the pcDNA5/FRT vector and have identified one potential second-site integrant after screening over 200 clones. DNA integrations can be detected by Southern blot. A single integrant will display a single band; double: two; triple: three, etc. We have maintained a number of Flp-In expression cell lines for over four months and have not observed any loss of the Flp-In expression construct, whether hygromycin selection was maintained or not.

When should I consider reversible integration (Flp-In system) vs irreversible integration (Jump-In system)?

Use irreversible integration (Jump-In system) if the transgene should be sustained in the mammalian genome for a long time. Use reversible integration such as Flp-In system if the transgene needs to be replaced with another gene of interest after a short period of time.

What is the difference between Jump-In and Flp-In systems?

The Jump-In system is PhiC31-integrase mediated and is a stable, targeted, and irreversible mammalian expression system, involving one integration step. The Jump-In TI (Targeted Integration) system needs two integration steps, both of which are targeted and irreversible. In contrast, the Flp-In system is a stable, targeted mammalian expression system that is reversible. The first integration is random (integration of pFRT/lacZeo) and the second integration (integration of the Flp-In expression vector) is targeted but reversible.

In the Flp-In system, how can one explain the presence of hygromycin-resistant clones that are also resistant to Zeocin antibiotic and are lacZ positive?

We have observed in-house that in cells where the FRT site has integrated into a very transcriptionally active locus in the host cell genome (seen more commonly in Flp-In CHO and Flp-In 293 cells but can also happen in Flp-In 3T3 cells and any other Flp-In host cell line), there is some "read-through" transcription and translation of the lacZ-Zeocin ORF post Flp-In recombination, even though the lacZ-Zeocin ORF does not have a bona fide promoter and ATG. In such cases, the hygromycin-resistant clones would also be lacZ positive and Zeocinantibiotic-resistant. To make sure that the integration is FRT site-specific and not random, we recommend doing a parallel control transfection with no pOG44 present. This should yield no surviving clones upon hygromycin selection, indicating that all the hygromycin-resistant clones obtained in the presence of pOG44 are indeed Flp recombinase-dependent and hence have the gene of interest integrated at the FRT site. Also, a Southern blot analysis of these clones will help verify that they do indeed have proper FRT integration of the gene of interest despite the expression of lacZ (although this is typically not necessary). Post Flp-Inrecombination, as long as you see hygromycin-resistant clones, we recommend that you select them and assay them for expression of your gene of interest.

I sequenced one of your vectors after PCR amplification and observed a difference from what is provided online (or in the manual). Should I be concerned?

Our vectors have not been completely sequenced. Your sequence data may differ when compared to what is provided. Known mutations that do not affect the function of the vector are annotated in public databases.

Are your vectors routinely sequenced?

No, our vectors are not routinely sequenced. Quality control and release criteria utilize other methods.

How was the reference sequence for your vectors created?

Sequences provided for our vectors have been compiled from information in sequence databases, published sequences, and other sources.


For Research Use Only. Not for use in diagnostic procedures.