Learn More
Invitrogen™ pEXP3-DEST Vector Kit
Catalog No. V96003
Encompass
Change view
Click to view available options
Antibiotic Resistance Bacterial:
Ampicillin (AmpR)
Catalog No. Antibiotic Resistance Bacterial
V96003 Ampicillin (AmpR)
1 options
Catalog No. V96003 Supplier Invitrogen™ Supplier No. V96003
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Includes
E. coli slyD- Extract, 2.5X IVPS E. coli Reaction Buffer, T7 Enzyme Mix, the Feed Buffer, 19 amino acid mix, Methionine, a control plasmid, DNAse/RNAse-free Water, and a Lumio Green Detection Kit for 20 real-time reactions or 100 in-gel applications. In addition to these components, the N-term Expressway Lumio kit also includes 6μg of the pEXP3-DEST vector and the C-term Expressway Lumio kit 6μg of the pEXP4-DEST vector.
Lumio™ detection reagent binds Lumio recognition sequence, small, six amino acid sequence (Cys-Cys-Pro-Gly-Cys-Cys)
  • This recognition sequence has high specificity and affinity, resulting in bright fluorescent signal for real-time protein production analysis and immediate in-gel protein detection
  • Specialized E. coli lysate, derived from slyD mutant, eliminates nonspecific binding of LumioGreen Detection Reagent to endogenous SlyD protein and provides optimal background for detection of recombinant proteins
  • LumioGreen Detection Kit is included in system for real-time detection of protein synthesis as well as easy in-gel protein detection
  • Lumio vectors also feature attR sites for efficient recombination with any attL-flanked Gateway entry vector
  • Vectors also feature N-terminal Lumio tag (pEXP3-DEST vector) with TEV cleavage site for efficient removal of Lumio sequence following purification
Cell Free Protein Expression for Functional Analysis, Cell-Free (in vitro) Expression, Cloning, Gateway Cloning, Protein Expression, Proteins, Expression, Isolation and Analysis
Specifications
Promoter T7
Product Type Gateway System Destination Expression Vector
Selection Agent (Eukaryotic) None
Content And Storage The Expressway™ Lumio™ Expression and Detection System includes the E. coli slyD- Extract, 2.5X IVPS E. coli Reaction Buffer, T7 Enzyme Mix, the Feed Buffer, 19 amino acid mix, Methionine, a control plasmid, DNAse/RNAse-free Water, and a Lumio™ Green Detection Kit for 20 real-time reactions or 100 in-gel applications. In addition to these components, the N-term Expressway™ Lumio™ kit also includes 6 μg of the pEXP3-DEST vector and the C-term Expressway™ Lumio™ kit 6 μg of the pEXP4-DEST vector. Store the vectors and the detection reagent at -20°C. Store all other components at -80°C. Guaranteed stable for 6 months when properly stored.
Antibiotic Resistance Bacterial Ampicillin (AmpR)
Cleavage TEV Protease Recognition Site
Protein Tag Lumio
Cloning Method Gateway
Vector pEXP, pDEST
Product Line Expressway, Gateway
For Research Use Only. Not for use in diagnostic procedures.