Learn More
Invitrogen™ E-Gel™ EX Agarose Gels
Invitrogen E-Gel EX precast agarose gels are designed for ultrasensitive and convenient DNA and RNA sample electrophoresis in as little as 10 to 20 minutes.
Supplier: Invitrogen™ G402021
Requires : Requires the E-Gel iBase™ Power System for running and the E-Gel Safe Imager™ Real-Time Transilluminator for real-time viewing of sample migration.
Description
Invitrogen E-Gel EX precast agarose gels are designed for ultrasensitive and convenient DNA and RNA sample electrophoresis in as little as 10 to 20 minutes. Each E-Gel agarose gel cassette contains all components and stain required for efficient gel separation and analysis–just load your samples and run. E-Gel agarose gels are ideal for analyzing PCR products, restriction digests, and plasmid preparations, or for DNA fragment library analysis. E-Gel agarose gels are also suitable for a quick check of RNA samples.
Features of E-Gel EX agarose gels include:
- Instantaneous setup—precast E-Gel agarose gels are ready for use when you are, and do not require gel preparation or additional buffers
- High sensitivity—detect 1 ng of sample DNA
- Rapid analysis—complete DNA sample separation in 10–20 minutes
- In-process control—view DNA sample migration in real time
- Easy sample access—openable cassette design allows easy in-gel access for convenient DNA sample excision or transfer to membranes
- For DNA and RNA—ideal for DNA sample analysis and suitable for quick check of RNA samples
Ultra-sensitive detection of DNA
E-Gel EX gels pre-stained with SYBR Gold DNA stain were developed for ultimate sensitivity and demonstrate up to 5-fold greater sensitivity than comparable gels containing ethidium bromide. This superior sensitivity allows you to use lower amounts of sample, saving time and money.
Fast and convenient analysis
E-Gel EX gels are our fastest-resolving agarose gels, enabling sample analysis in as little as 10–20 minutes. Each E-Gel agarose gel contains agarose, electrodes, SYBR Gold DNA stain, and a bufferless ion exchange system packaged inside a dry disposable cassette. E-Gel technology does not required any gel or buffer preparation or gel straining steps. Just load your samples and run.
Live monitoring of DNA migration
The E-Gel EX agarose gels are pre-stained with SYBR Gold II DNA gel stain with an excitation wavelength in the blue-light spectrum. When used on the E-Gel Power Snap Electrophoresis Device with integrated blue-light trans-illuminator, safe real-time monitoring of DNA or RNA migration is enabled without risk to eyes or possible damage of the sample, unlike UV illumination.
Quick check of RNA samples
Besides DNA analysis, E-Gel EX gels can also be used for fast analysis of RNA samples to check their integrity before proceeding with downstream applications.
Easy access to the gel
E-Gel cassettes are designed for convenient opening with a Gel Knife (Cat. No. EI9010), for easy excision of specific bands or transfer of the gel to a membrane for southern blot analysis.
Running and imaging devices
E-Gel EX gels require the Invitrogen E-Gel Power Snap Electrophoresis System for gel running and viewing. The system consists of an electrophoresis device and a camera for fast and convenient E-Gel agarose gel separation and analysis.
Available in a variety of gel formats
E-Gel agarose gels are available in variety of gel percentage, stain, and well formats. The 11-well E-Gel EX agarose gels are available in 1%, 2%, and 4% gel percentages.
Order Info
Shipping Condition: Room Temperature
Chemical Identifiers
| E-Gel | |
| 20 μL | |
| Molecular Weight | |
| Single Comb |
| SYBR Gold | |
| 11-well | |
| Room Temperature | |
| EX Agarose Gel |
Specifications
| E-Gel™ 1 Kb Plus Express Ladder, Millennium™ RNA Markers | |
| 1% | |
| SYBR Gold | |
| 100 to 5000 bp | |
| Molecular Weight | |
| Preparative without dyes, Non-preparative | |
| Room Temperature | |
| EX Agarose Gel |
| • E-Gel™ EX 20-Paks contain 20 gels Store gels at room temperature. Guaranteed stable for 3 months when properly stored. |
|
| E-Gel | |
| 20 μL | |
| 11-well | |
| 20 Gels/Pk. | |
| Double-Stranded DNA (dsDNA), Single-Stranded RNA, Circular RNA | |
| Single Comb |
For Research Use Only. Not for use in diagnostic procedures.