missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MegaX DH10B T1R Electrocomp™ Cells

Catalog No. C640003
Encompass
Change view
Click to view available options
Quantity:
5 x 100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C640003 5 x 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C640003 Supplier Invitrogen™ Supplier No. C640003

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Contains: MegaX DH10B™ T1R Electrocomp™ C: 5 vials, 100 μL each; pUC19 DNA (10pg/μL): 1 vial, 50μL; Recovery Medium: 3 bottles, 10mL each

MegaX DH10B T1R Electrocomp Cells are the highest-efficiency electrocompetent cells available. Improved production processes provide a transformation efficiency reaching >3 x 1010 cfu/μg of pUC DNA.

MegaX DH10B T1R Electrocomp Cells are the highest-efficiency electrocompetent cells available. Improved production processes provide a transformation efficiency reaching >3 x 1010 cfu/μg of pUC DNA, making MegaX DH10B T1R cells ideal for highly demanding cloning or cDNA and gDNA library construction applications.

The DH10B T1R strain has all the benefits of DH10B genotype, with the addition of the tonA (also known as fhuA) genotype that confers resistance to T1 and T5 phage infection. The inclusion of the tonA genotype offers added security to protect valuable clones and libraries. MegaX DH10B T1R cells carry mutations in the methylation-dependent E. coli K12 restriction system (mcrA, mcrBC, and mrr) allowing construction of genomic libraries of both prokaryotic and eukaryotic genomic DNA, as well as efficient plasmid rescue from eukaryotic genomes. This strain also has the lacZΔM15 genotype, providing for the option of blue-white screening on plates containing either X-Gal or Bluo-Gal. Finally, the endA1 mutation makes DH10B T1R an excellent vehicle for amplifying plasmid DNA for subsequent extraction and purification.

MegaX DH10B T1R Electrocomp Cells offer:
• Highest transformation efficiency of >3 x 1010 cfu/μg
• tonA (fhuA) genotype to confer resistance to T1 and T5 phage
• lacZΔM15 marker provides α-complementation of the β-galactosidase gene allowing blue/white screening on agar plates containing X-Gal or Bluo-Gal
• mcrA genotypic marker and the mcrBC, mrr deletion allows for cloning DNA that contains methylcytosine and methyladenine
• endA1 mutation enables increased plasmid yield and quantity
• recA1 for reduced occurrence of nonspecific recombination in cloned DNA

Note: A high-voltage electroporation apparatus is required.

Genotype
F–mcrA Δ(mrr-hsdRMS-mcrBC) φ80lacZΔM15 ΔlacX74 recA1 endA1 araD139 Δ(ara-leu)7697 galU galK λrpsL(StrR) nupG tonA

Find the strain and format that fits your needs
• We offer other DH strains in chemically competent and electrocompetent cell formats to meet your specific needs.
• Electroporation cuvettes are available in 0.1 cm (Cat. No.P41050), 0.2 cm (Cat. No. P45050), and 0.4 cm (Cat. No. P46050) gaps.
• Strains are available in several MultiShot formats for high throughput applications.

Order Info

Shipping Condition: Dry Ice

Specifications

Product Type Electrocompetent Cells
Contains F' Episome No
Improves Plasmid Quality Yes (endA1)
Cloning Methylated DNA Yes (mcrA)
Transformation Efficiency Level High Efficiency (>3 x 1010 cfu/μg)
Content And Storage • MegaX DH10B T1R Electrocomp Cells (5 x 100 μL)
Store Competent Cells at –80°C.

• pUC19 DNA (50 μL)
Store pUC19 DNA at –20°C.

• Recovery Medium (3 x 10 mL)
Store Recovery Medium at 4°C
Antibiotic Resistance Bacterial Yes (Streptomycin)
Cloning Unstable DNA Not suitable for cloning unstable DNA
Blue/White Screening Yes (lacZΔM15)
High-throughput Compatibility Low
Plasmid May be used for plasmids >100 kb
Preparing Unmethylated DNA No
Reduces Recombination Yes (recA1)
Shipping Condition Dry Ice
T1 Phage - Resistant (tonA) Yes
Species E. coli (K12)
Format Tube
Product Line Electrocomp
Quantity 5 x 100 μL
Show More Show Less
I'm making a yeast genomic library and want to transform and amplify it in one of your competent cell strains. What genotype features should I look for in choosing a good strain?

We would recommend a mcr/mrr- strain, which prevents restriction of methylated eukaryotic DNA in the E. coli host. We would also recommend using a T1R strain, as T1 is a common contaminant in genomic/cDNA libraries.

For Research Use Only. Not for use in diagnostic procedures.