missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DDM (n-dodecyl β-D-maltoside) (10%)

Catalog No. BN2005
Encompass
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BN2005 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BN2005 Supplier Invitrogen™ Supplier No. BN2005

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Detergent solution used to prepare native protein samples for use with the NativePAGE™ Gel System

  • Ready-to-use detergent solution improves solubility of hydrophobic and membrane proteins during sample preparation
  • The samples prepared with 10% DDM, 5% Digitonin, or the NativePAGE Sample Prep Kit are compatible with NativePAGE Novex Bis-Tris Gels for native electrophoresis showing increased resolution and reduced streaking

1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Wet Ice

Specifications

Content And Storage 1 ml of solution. Store at +4°C.
Form Liquid
Quantity 1 mL
Reagent Type DDM
Shipping Condition Wet Ice
Product Type DDM
Can I use the NativePAGE Sample and Running buffers with NuPAGE Tris-Acetate gels for native applications?

NativePAGE Sample Buffers and Running buffers were developed specifically for use with the NativePAGE Bis-Tris gels. We do not recommend using these buffers for native applications with any other gels, including NuPAGE Tris-Acetate gels. When running Tris-Acetate gels under native conditions, we recommend using the Invitrogen Tris-Glycine Native Sample and running buffers.

For Research Use Only. Not for use in diagnostic procedures.