missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ siPORT™ Amine Transfection Agent with manual

Catalog No. AM4503M
Encompass
Change view
Click to view available options
Quantity:
400 μL
1 mL
Product Type:
Transfection Reagent
Transfection Reagent with Manual
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
AM4503M 1 mL Transfection Reagent with Manual
AM4502 400 μL Transfection Reagent
AM4503 1 mL Transfection Reagent
AM4502M 400 μL Transfection Reagent with Manual
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AM4503M Supplier Invitrogen™ Supplier No. AM4503M

missing translation for 'validMainframeMsg'

Includes

Supplied in one tube containing 0.4mL.

Animal origin-free polyamine-based transfection agent for use in transfecting cells during subculturing—saving time, without increasing toxicity

  • Successful siRNA experiment requires the appropriate transfection agent for your cells, and one that is optimized for siRNA delivery
  • Efficiency with which different transfection reagents deliver nucleic acids can vary dramatically with the type of nucleic acid being transfected (RNA vs. DNA, single-stranded vs. double-stranded) and the cells being used

Accessory Products:

  • siPORT™ NeoFX™ Transfection Agent (Mfr. Nos. AM4511 and AM4510), a cationic and neutral lipid mixture, is also available
  • For testing which transfection agent is best with your system, the Silencer™ siRNA Transfection II Kit (Mfr. No. AM1631) which contains the two different transfection agents and controls for transfection optimization, is recommended

Cell Culture, RNAi Transfection, Synthetic siRNA Transfection, Transfection

Order Info

Shipping Condition: Wet Ice

Specifications

Content And Storage Store at 4°C.
Cell Type Primary Cells
Format 6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks
Sample Type Synthetic siRNA
Transfection Technique Lipid-based Transfection
For Use With (Application) Transfection
High-throughput Compatibility Not High-throughput Compatible (Manual)
Product Line Ambion, siPORT
Product Type Transfection Reagent with Manual
Quantity 1 mL
Serum Compatible Yes
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.