missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ siPORT™ Amine Transfection Agent with manual

Animal origin-free polyamine-based transfection agent for use in transfecting cells during subculturing—saving time, without increasing toxicity

Supplier:  Invitrogen™ AM4502M

Encompass

 View more versions of this product

Catalog No. AM4502M


Only null left

Includes: Supplied in one tube containing 0.4mL.

Description

Description

  • Successful siRNA experiment requires the appropriate transfection agent for your cells, and one that is optimized for siRNA delivery
  • Efficiency with which different transfection reagents deliver nucleic acids can vary dramatically with the type of nucleic acid being transfected (RNA vs. DNA, single-stranded vs. double-stranded) and the cells being used

Accessory Products:

  • siPORT™ NeoFX™ Transfection Agent (Mfr. Nos. AM4511 and AM4510), a cationic and neutral lipid mixture, is also available
  • For testing which transfection agent is best with your system, the Silencer™ siRNA Transfection II Kit (Mfr. No. AM1631) which contains the two different transfection agents and controls for transfection optimization, is recommended

Cell Culture, RNAi Transfection, Synthetic siRNA Transfection, Transfection

Order Info

Shipping Condition: Wet Ice

Specifications

Chemical Identifiers

Store at 4°C.
6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks
Lipid-based Transfection
Not High-throughput Compatible (Manual)
Yes
Primary Cells
Synthetic siRNA
Transfection
Ambion, siPORT

Specifications

Store at 4°C.
6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks
Lipid-based Transfection
Not High-throughput Compatible (Manual)
Transfection Reagent with Manual
Yes
Primary Cells
Synthetic siRNA
Transfection
Ambion, siPORT
400 μL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.