Learn More
Invertebrate Set
Enhance your slide collection with this fine survey set.
Supplier:  Customer Channels Group (Ccg) S10034
Catalog No. S10034
Only null left
Description
Description
Set of 25 prepared slides presents a survey of organisms. Seven of the slides show protozoans including Amoeba, Paramecium, and Stentor. The other slides from the invertebrate group include sponges; cniderians; flat, round and segmented worms; bryozoa; mollusca; arthropoda; and echinodermata.
Includes:
  • Amoeba proteus, w.m.
  • Clam gill, c.s.
  • Daphnia, w.m.
  • Dog flea, w.m.
  • Earthworm, c.s. of posterior
  • Euglena, w.m.
  • Fasciola hepatica (liver fluke), w.m.
  • Hydra budding, w.m.
  • Obelia, hydroid colony, w.m.
  • Paramecium caudatum, w.m.
  • Paramecium in fission, w.m.
  • Pectinatella (Freshwater Bryozoan), w.m.
  • Planaria injected, w.m.
  • Radiolarian ooze, w.m.
  • Rotifer, w.m.
  • Scypha, c.s. and l.s.
  • Sponge spicules, w.m.
  • Starfish, w.m.
  • Stentor, w.m.
  • Tapeworm, scolex and immature segments, w.m.
  • Tapeworm, mature segment, w.m.
  • Tapeworm, gravid segment, w.m.
  • Trichinella spiralis, larvae in muscle, sec.
  • Trypanosoma rhodesiense parasitic blood smear
Specifications
Specifications
Invertebrates
25
Product Suggestions
Product Suggestions