missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Intermediate Polar Fused Silica Tubing

Catalog No. 111004962
Change view
Click to view available options
Overall Length (Metric):
1 m
5 m
15 m
60 m
Diameter (English) Inner:
0.25 mm
0.32 mm
0.53 mm
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Overall Length (Metric) Diameter (English) Inner
111004962 5 m 0.25 mm
111020813 15 m 0.25 mm
111003597 60 m 0.25 mm
111004860 1 m 0.32 mm
111004673 5 m 0.53 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 111004962 Supplier MilliporeSigma™ Supelco™ Supplier No. 25747

Ideal for use with injection solvents such as acetone, methylene chloride (dichloromethane), and toluene

  • Use as transfer lines, guard columns, or retention gaps
  • Can be used to make custom columns
  • Ideal for use with injection solvents such as acetone, methylene chloride (dichloromethane), and toluene
  • Composition: Phenyl/Methyl
  • Deactivated

Specifications

For Use With Injection Solvents Such as Acetone, Methylene Chloride (Dichloromethane), and Toluene
Type Tubing
For Use With (Application) Use as Transfer Lines, Guard Columns, Retention Gaps, or Custom Columns
Diameter (English) Inner 0.25 mm
Overall Length (Metric) 5 m
Material Silica