missing translation for 'onlineSavingsMsg'
Learn More

Interferents A For ICP/MS, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Supplier:  SPEX CertiPrep Group LLC CLINTA1

Catalog No. CL-INT-A1


Only null left

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Cl-21,215μg/mL
Ca3000μg/mL
Fe2500μg/mL
Na2500μg/mL
C2000μg/mL
Al1000μg/mL
K1000μg/mL
Mg1000μg/mL
P1000μg/mL
S1000μg/mL
Mo20μg/mL
Ti20μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Interferents A1
Triple-leached LDPE bottle
GC-MS
Certified Reference Material
125 mL
Components in 5% HNO3/tr. HF:Al    •1000μg/mLCa    •3000μg/mLC    •2000μg/mLCl    •21, 215μg/mLFe    •2500μg/mLMg    •1000μg/mLMo    •20μg/mLP    •1000μg/mLK    •1
For CLP-M
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Nitric Acid/Trace Hydrogen Fluoride
ICP/ICP-MS
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

Product intended for laboratory use only