missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Interference Check Standard 18 without Mercury For Environmental Analysis, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Supplier: SPEX CertiPrep Group LLC INTER18500N
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| K | 20,000μg/mL |
| As | 1000μg/mL |
| Pb | 1000μg/mL |
| Tl | 1000μg/mL |
| Se | 500μg/mL |
| Ag | 300μg/mL |
| Ba | 300μg/mL |
| Cd | 300μg/mL |
| Co | 300μg/mL |
| Cr | 300μg/mL |
| Cu | 300μg/mL |
| Ni | 300μg/mL |
| V | 300μg/mL |
| Zn | 300μg/mL |
| Mn | 200μg/mL |
| Be | 100μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
| Interference Check Std 18 without Mercury | |
| Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
| AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
| 5% HNO3 | |
| Environmental |
| Triple-leached LDPE bottle | |
| AA, ICP | |
| Certified Reference Material | |
| 500 mL | |
| Various |
Product intended for laboratory use only